Skip to Content
Merck

Skip To

C3993

Carvedilol

≥98% (HPLC), β-adrenergic blocker, solid

Synonym(s):

1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol, BM-14190

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H26N2O4
CAS Number:
Molecular Weight:
406.47
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Carvedilol, ≥98% (HPLC), solid

Quality Segment

assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

DMSO: >20 mg/mL

originator

GlaxoSmithKline

SMILES string

OC(COC1=CC=CC2=C1C3=C(C=CC=C3)N2)CNCCOC4=CC=CC=C4OC

InChI

1S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27H,13-16H2,1H3

InChI key

OGHNVEJMJSYVRP-UHFFFAOYSA-N

Application

Carvedilol has been used:
  • as a β blocker to determine its effect on hypoxia inducible factor (HIF)-mediated erythropoiesis under hypoxia in vivo
  • as a βAR antagonist for inhibition of bARs
  • as a β-blocker to examine airway mechanics in fungus-challenged mice

Biochem/physiol Actions

Cavedilol is a non-selective β-adrenergic blocker with α1 blocking activity.
Cavedilol is a non-selective β-adrenergic blocker with α1 blocking activity. Carvedilol is used specifically for the treatment of heart failure and high blood pressure. It has been shown to improve left ventricular ejection fraction and may reduce mortality.

Features and Benefits

This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1096622Y0001426Y0001427
form

solid

form

-

form

-

form

-

assay

≥98% (HPLC)

assay

-

assay

-

assay

-

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

solubility

DMSO: >20 mg/mL

solubility

-

solubility

-

solubility

-

originator

GlaxoSmithKline

originator

-

originator

-

originator

-

color

white to off-white

color

-

color

-

color

-


Still not finding the right product?

Explore all of our products under Carvedilol


pictograms

Health hazardEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2

target_organs

Liver,spleen,Uterus (including cervix),Adrenal gland

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
C3993-50MG04061833486801
C3993-250MG04061832081571

Questions

Reviews

No rating value

Active Filters