Skip to Content
Merck

Skip To

C0832

Clozapine N-oxide

≥98% (HPLC), solid, 5-HT₂ serotonin receptor antagonist

Synonym(s):

8-Chloro-11-(4-methyl-1-piperazinyl)-5H-dibenzo[b,e](1,4)diazepine N-oxide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C18H19ClN4O
CAS Number:
Molecular Weight:
342.82
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Form:
solid
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Clozapine N-oxide,

form

solid

Quality Level

SMILES string

C[N+]1([O-])CCN(CC1)C2=Nc3cc(Cl)ccc3Nc4ccccc24

InChI

1S/C18H19ClN4O/c1-23(24)10-8-22(9-11-23)18-14-4-2-3-5-15(14)20-16-7-6-13(19)12-17(16)21-18/h2-7,12,20H,8-11H2,1H3

InChI key

OGUCZBIQSYYWEF-UHFFFAOYSA-N

General description

Clozapine N-oxide (CNO), inactive from of clozapine drug, activates G-protein-coupled receptors (GPCRs). CNO serves as a synthetic ligand for engineered human muscarinic receptor. It elicits clozapine-like effects upon metabolic conversion. Clozapine conversion to CNO favors its passage through blood-brain barrier.

Application

Clozapine N-oxide has been used:
  • for the activation of human M4 muscarinic (hM4Di) in primary motor cortex
  • for inducing in vivo chemogenetic manipulation and impairing locomotor activity in Th-cre transgenic mice
  • in behavioral testing in mice and for stimulating (HEK293) cells

Biochem/physiol Actions

5-HT2 serotonin receptor antagonist; major metabolite of clopazine that can be monitored by HPLC.
5-HT2 serotonin receptor antagonist; major metabolite of clopazine.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2304C6305D5676
form

solid

form

powder

form

powder

form

powder

Quality Level

200

Quality Level

-

Quality Level

300

Quality Level

200


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



The DREADD agonist clozapine N-oxide (CNO) is reverse-metabolized to clozapine and produces clozapine-like interoceptive stimulus effects in rats and mice
Manvich DF, et al.
Scientific Reports, 8(1), 3840-3840 (2018)
Noelle C Anastasio et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 44(10), 1752-1761 (2019-04-20)
Food intake is essential for survival, but maladaptive patterns of intake, possibly encoded by a preexisting vulnerability coupled with the influence of environmental variables, can modify the reward value of food. Impulsivity, a predisposition toward rapid unplanned reactions to stimuli
Chemogenetic and optogenetic activation of galphas signaling in the basolateral amygdala induces acute and social anxiety-like states
Siuda ER, et al.
Neuropsychopharmacology, 41(8), 2011-2011 (2016)



Global Trade Item Number

SKUGTIN
C0832-5MG04061838445520

Questions

Reviews

No rating value

Active Filters