Skip to Content
Merck

Skip To

B8931

Magenta-Gal

reagent for selection of recombinant bacterial clones

Synonym(s):

Red-Gal, 5-Bromo-6-chloro-3-indolyl β-D-galactopyranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C14H15BrClNO6
CAS Number:
Molecular Weight:
408.63
NACRES:
NA.85
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

sterility

non-sterile

product line

BioReagent

assay

≥98.0% (HPLC)

form

powder

solubility

methanol: soluble

suitability

suitable for substrate for β-galactosidase

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@H](Oc2c[nH]c3cc(Cl)c(Br)cc23)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H15BrClNO6/c15-6-1-5-8(2-7(6)16)17-3-9(5)22-14-13(21)12(20)11(19)10(4-18)23-14/h1-3,10-14,17-21H,4H2/t10-,11+,12+,13-,14+/m1/s1

InChI key

CHRVKCMQIZYLNM-HTOAHKCRSA-N

General description

Red-Gal is a chromogenic substrate for β-galactosidase, used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Red-Gal is designed to replace X-Gal in blue-white selection of recombinant bacterial colonies with the lac+ phenotype.
Red-Gal is a chromogenic substrate for b-galactosidase,[1] used to determine the presence or absence of a cloned DNA insert in bacteria growing on agar plates. Red-Gal is designed to replace X-Gal in the blue-white selection of recombinant bacterial colonies with the lac+ phenotype.

Application

Chromogenic substrate for β-galactosidase producing a red or magenta insoluble product.
Magenta-Gal has been used for staining synthetic cytoplasmic or nuclear LacZ mRNA. It has also been used to detect the expression of the EphB1-β-gal fusion protein.[1] ,Magenta-Gal is suitable for use in the selection of recombinant bacterial colonies with the lac+ phenotype.

Biochem/physiol Actions

When Red-Gal is hydrolyzed by β-galactosidase, the resulting product will form a red precipitate. Lac+ colonies grown in the presence of Red-Gal turn an intense red color, allowing for easy differentiation between lac+ and lac- colonies.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S9811B2904RES1364C
suitability

suitable for substrate for β-galactosidase

suitability

suitable for β-galactosidase test

suitability

suitable for substrate for β-galactosidase

suitability

-

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

grade

-

solubility

methanol: soluble

solubility

DMF: 50 mg/mL

solubility

DMF: soluble

solubility

-

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

room temp

storage temp.

−20°C

storage temp.

−20°C

assay

≥98.0% (HPLC)

assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

-


Still not finding the right product?

Explore all of our products under Magenta-Gal


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
B8931-25MG04061833074022

Questions

Reviews

No rating value

Active Filters