Skip to Content
Merck

Skip To

B6185

Bionic Buffer, 10X Concentrate

for Northern and Southern blotting, solution

Synonym(s):

Sodium Borate Electrophoresis Buffer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
41105319
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

description

Mode of use Voltage of 150 V is recommended for small electrophoresis chambers while a mid to large apparatus may be run at 200-250 V. User should test their chamber to determine optimum voltage.

form

solution

concentration

10 ×

impurities

DNase, RNase and nickase, none detected

pH

8.0-8.5 (20-25 °C)

storage temp.

room temp

General description

Bionic Buffer is a unique alternative to traditional TBE (TRIS borate-EDTA) and TAE (TRIS acetate-EDTA) electrophoresis buffers. Bionic Buffer allows for higher voltage runs, extremely rapid running times, and high-resolution band separation.

Application

Suitable for:
  • Use as running buffer for nucleic acid electrophoresis, which may be part of Northern and Southern blot hybridization procedures
  • Use with TBA and TAE gels
  • Preparation of agarose gels

Features and Benefits

  • Faster run times
  • Sharper bands
  • Easy to dilute and use
  • Flexible use with a variety of gel types

Packaging

The 4 L and 10 L sizes are supplied in dispensers with a spigot.

Legal Information

Bionic is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Test electrophoresis chambers to determine highest voltage. Voltages beyond the recommended range for your apparatus may result in blown fuses or damage to your apparatus.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B6429B493093290
grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

grade

Molecular Biology

concentration

10 ×

concentration

-

concentration

-

concentration

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

solution

form

solution

form

liquid

form

solution

pH

8.0-8.5 (20-25 °C)

pH

-

pH

-

pH

8.6-9.0

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters