Skip to Content
Merck

Skip To

A9441

Aflatoxin B + G mixture

dry concentrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.77
UNSPSC Code:
12352200
Form:
dry concentrate
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

dry concentrate

Quality Segment

storage temp.

2-8°C

General description

Aflatoxins are hepatotoxic metabolites, which are produced by molds, such as Aspergillus flavus and Aspergillus parasiticus. They are naturally occurring poisonous carcinogens and the most widely known mycotoxins. They mainly contaminate fruits, vegetables and grains. Aflatoxins contaminate human food and animal feed. They are associated with liver cancer, immune system suppression, reduced growth rates, losses in feed efficiency and death. Aflatoxin B is more carcinogenic than Aflatoxin G.

Application

Aflatoxin B + G mixture has been used for the quantification of aflatoxin by high-performance thin-layer chromatography, using a charge-coupled device detector.

Preparation Note

Concentration: 5 μg aflatoxin B1, 1.5 μg aflatoxin B2, 5 μg aflatoxin G1, 1.5 μg aflatoxin G2 per mL after reconstitution to approx. 5 ml.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A0138A6655A0263
form

dry concentrate

form

powder

form

powder

form

powder

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Carc. 1B - Muta. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
A9441-1VL04061832938875

Questions

Reviews

No rating value

Active Filters