Skip to Content
Merck

Skip To

A9393

Ampicillin

anhydrous, 96.0-102.0% (anhydrous basis)

Synonym(s):

(-)-6-(2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, Ampicillin acid, Ampicillin anhydrous, D-(−)-α-Aminobenzylpenicillin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C16H19N3O4S
CAS Number:
Molecular Weight:
349.40
UNSPSC Code:
51281703
NACRES:
NA.85
PubChem Substance ID:
EC Number:
200-709-7
Beilstein/REAXYS Number:
1090925
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

96.0-102.0% (anhydrous basis)

form

solid

pKa (25 °C)

2.5 (COOH), 7.3 (NH2)

mp

208 °C (dec.) (lit.)

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)[C@H](N)c3ccccc3)C(O)=O

InChI

1S/C16H19N3O4S/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23)/t9-,10-,11+,14-/m1/s1

InChI key

AVKUERGKIZMTKX-NJBDSQKTSA-N

General description

Chemical structure: β-lactam

Application

Ampicillin has been used to study antibiotic resistance and penetration limitations, the synergy between multiple antibiotics, certain bloodstream infections, and has been used to develop PCR assays to detect resistance genes in cerebrospinal fluid.[1][2][3][4]

Biochem/physiol Actions

Mode of Action: Ampicillin is a semisynthetic penicillin and a β-lactam antibiotic that inhibits bacterial cell-wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane.

Mode of Resistance: Administration with β-lactamase cleaves the β-lactam ring of Ampicillin and inactivates it.

Antimicrobial Spectrum: Effective against both Gram-positive (similar to benzylpenicillin) and Gram-negative bacteria (similar to tetracyclines and chloramphenicol.
β-lactams are inactivated by β-lactamases and for this reason ampicillin is used with a β-lactamase inhibitor.[5][6]

Preparation Note

Ampicillin is reported as slightly soluble in water, practically insoluble in alcohol, chloroform, ether and fixed oils but soluble in dilute acids or bases. The solution should not be autoclaved; a stock solution should be sterilized through filtration and stored frozen, where it will be stable for months.

Other Notes

Store under argon. Keep container tightly closed in a dry and well-ventilated place, hygroscopic.

Disclaimer

This product has been reported stable as supplied at 25°C at 43% and 81% relative humidity for six weeks. Additional studies have shown that the stability of Ampicillin in solution is a function of pH, temperature and the identity of the buffer. It′s activity is quickly lost when stored above pH 7. Optimal storage conditions are suggested as 2-8°C, and pH 3.8-5 where its activity was retained at 90%+ for a week.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A159359349A1000000
antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

-

antibiotic activity spectrum

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

-

assay

96.0-102.0% (anhydrous basis)

assay

-

assay

96.0-102.0% anhydrous basis

assay

-

form

solid

form

solid

form

-

form

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

mp

208 °C (dec.) (lit.)

mp

198-200 °C (dec.) (lit.)

mp

208 °C (dec.) (lit.)

mp

208 °C (dec.) (lit.)


Still not finding the right product?

Explore all of our products under Ampicillin


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
A9393-5G04061835505883
A9393-100G04061833689677
A9393-25G04061835515752

Questions

1–7 of 7 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Which Ampicillin product has higher water solubility, Product A9393 or Product A9518?

    1 answer
    1. Product No. A9518 (Ampicillin, sodium salt) is soluble at 50 mg/mL in water.

      Helpful?

  4. How do I use Ampicllin in agar plates?

    1 answer
    1. For plates, add ampicillin to autoclaved media cooled to 48°C, to a final concentration of 35-50 μg/mL. Pour the plates, and allow them to cool completely. Plates may be stored at 2-8°C for up to only 1-2 weeks before use.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. What is the solubility of Product A9393, Ampicillin?

    1 answer
    1. The solubility of ampicillin is tested at 50 mg/mL in 1M ammonium hydroxide. Additional solubility information can be found on the product information sheet (under Documents, above).

      Helpful?

  7. Is Ampicillin product stable in solution?

    1 answer
    1. We recommend using freshly prepared solutions. A concentrated solution of sodium ampicillin in water (approximately 330 mg/mL) may be expected to lose about 10% potency in 20 hours at -20°C.

      Helpful?

Reviews

No rating value

Active Filters