Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Form:
lyophilized powder
Assay:
≥98% (SDS-PAGE)
Biological source:
egg white
Mol wt:
glycoprotein 66 kDa, subunit 16 kDa
Pricing and availability is not currently available.
biological source
egg white
Quality Level
product line
BioUltra
assay
≥98% (SDS-PAGE)
form
lyophilized powder
specific activity
≥10 units/mg protein (E1%/280)
mol wt
glycoprotein 66 kDa, subunit 16 kDa
storage temp.
2-8°C
General description
Avidin protein is a homotetrameric protein (68kDa) that is obtained from egg whites. This egg protein binds strongly to biotin. Thus, avidin-biotin association has been utilized in immunoassays to detect the localization of antigens in tissues[1]. The use of avidin-biotin immunoassay enhances the sensitivity of the technique and facilitates the detection of antigens in low quantities.
Application
Avidin egg white was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding. Egg white was used at 80 nmol.
Preparation Note
Affinity purified
Other Notes
One unit will bind 1.0 μg of d-biotin.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥98% (SDS-PAGE) | assay - | assay - | assay - |
| biological source egg white | biological source - | biological source egg white | biological source avidin from egg white, enzyme from horseradish |
| mol wt glycoprotein 66 kDa, subunit 16 kDa | mol wt - | mol wt - | mol wt - |
| form lyophilized powder | form lyophilized powder | form aqueous glycerol suspension | form lyophilized powder |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| specific activity ≥10 units/mg protein (E1%/280) | specific activity - | specific activity - | specific activity - |
Still not finding the right product?
Explore all of our products under Avidin from egg white
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Lee B Sims et al.
Investigative ophthalmology & visual science, 60(5), 1696-1705 (2019-04-23)
The goal of this work was to design and assess the ability of unmodified and surface-modified poly(lactic-co-glycolic acid) (PLGA) nanoparticles (NPs) to enhance cell association, provide efficacy in retinoblastoma cells, and overcome current administration challenges, including hydrolysis and precipitation, of
Roland Hager et al.
Scientific reports, 7, 42075-42075 (2017-02-16)
Arrays of individual molecules can combine the advantages of microarrays and single-molecule studies. They miniaturize assays to reduce sample and reagent consumption and increase throughput, and additionally uncover static and dynamic heterogeneity usually masked in molecular ensembles. However, realizing single-DNA
Lee B Sims et al.
Journal of nanobiotechnology, 15(1), 67-67 (2017-10-07)
Advanced stage cancer treatments are often invasive and painful-typically comprised of surgery, chemotherapy, and/or radiation treatment. Low transport efficiency during systemic chemotherapy may require high chemotherapeutic doses to effectively target cancerous tissue, resulting in systemic toxicity. Nanotherapeutic platforms have been
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A9275-25MG | 04061833401682 |
| A9275-2MG | 04061833401699 |
| A9275-100MG | 04061835553747 |
| A9275-10MG | 04061833401668 |
| A9275-5MG | 04061833401705 |
| A9275-1MG | 04061833401675 |



