Skip to Content
Merck

Skip To

A7627

L-Alanine

≥98% (TLC), suitable for cell culture

Synonym(s):

(S)-2-Aminopropionic acid, L-α-Aminopropionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
NACRES:
NA.26
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
200-273-8
MDL number:
Beilstein/REAXYS Number:
1720248
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Alanine, ≥98% (TLC)

Quality Level

assay

≥98% (TLC)

form

powder

technique(s)

cell culture | mammalian: suitable

color

, Colorless or White

solubility

H2O: 50 mg/mL, clear, colorless

application(s)

peptide synthesis

SMILES string

C[C@H](N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1

InChI key

QNAYBMKLOCPYGJ-REOHCLBHSA-N

Gene Information

human ... CA1(759), CA2(760)

General description

L-Alanine, a non-essential amino acid, is produced enzymatically from L-aspartate using aspartate β-decarboxylase. It is the smallest among all the aminoacids.

Application

L-Alanine has been used as a supplement in complete medium for cell culture.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A7469W38182905129
form

powder

form

powder

form

chunks, crystalline powder

form

powder or crystals

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

assay

≥98% (TLC)

assay

98.5-101.0%

assay

≥99%

assay

≥99.5% (NT)

color

, Colorless or White

color

white

color

-

color

colorless to white

solubility

H2O: 50 mg/mL, clear, colorless

solubility

H2O: 100 mg/mL

solubility

H2O: soluble 89.1 g/L at 20 °C (completely)

solubility

H2O: 1 M at 20 °C, clear, colorless

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

200


Still not finding the right product?

Explore all of our products under L-Alanine


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



The Prokaryotes: Vol. 6: Proteobacteria: Gamma Subclass, 2663-2679 (2006)
Berg JM
Biochemistry (2002)
An internal sensor detects dietary amino acids and promotes food consumption in Drosophila
Zhe Yang
Nature, 1-36 (2017)



Global Trade Item Number

SKUGTIN
A7627-1KG04061826210680
A7627-500G04061833385555
A7627-1G04061833385470
A7627-100G04061833385463

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters