Skip to Content
Merck

Skip To

A7219

L-Aspartic acid

from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%

Synonym(s):

(S)-(+)-Aminosuccinic acid, (S)-Aminobutanedioic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
HO2CCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
133.10
NACRES:
NA.75
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
200-291-6
MDL number:
Beilstein/REAXYS Number:
1723530
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

non-animal source

Quality Segment

agency

meets EP testing specifications, meets USP testing specifications

assay

98.5-101.0%

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

white to off-white

mp

>300 °C (dec.) (lit.)

solubility

1 M HCl: 100 mg/mL

SMILES string

N[C@@H](CC(O)=O)C(O)=O

InChI

1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1

InChI key

CKLJMWTZIZZHCS-REOHCLBHSA-N

Gene Information

human ... CA1(759), CA2(760)
rat ... Grin2a(24409)

General description

Aspartic acid(ASP)/aspartate, a non-essential amino acid, is naturally produced by mammals. ASP is one of the 20 building-block amino acids of proteins and has a one letter code, D. L-Aspartic acid can be manufactured industrially by an enzymatic process using the aspartase enzyme.

Application

L-Aspartic acid has been used:
  • as an aspartame (ASP) metabolite to determine acetylcholinesterase (AChE) activity in the hippocampal homogenate of suckling rats[1]
  • as printed chemotaxis ligands to screen chemotaxis receptors with high-throughput technologies and verify the interactions with surface plasmon resonance (SPR) and nuclear magnetic resonance (NMR)[2]
  • as a supplement in Dulbecco′s modified Eagle′s medium (DMEM) for supplementation assays of various cultured cell lines[3]

Biochem/physiol Actions

Principal neurotransmitter for fast synaptic excitation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9256A5474PHR1104
form

powder

form

powder, crystals or chunks

form

crystalline powder

form

-

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

cell culture | mammalian: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

assay

98.5-101.0%

assay

≥98% (HPLC)

assay

≥98.5%

assay

-

color

white to off-white

color

white to off-white

color

-

color

-

mp

>300 °C (dec.) (lit.)

mp

>300 °C (dec.) (lit.)

mp

>300 °C (dec.) (lit.)

mp

>300 °C (dec.) (lit.)

solubility

1 M HCl: 100 mg/mL

solubility

H2O: 5 mg/mL, 0.5 M HCl: 50 mg/mL (with heat), 1 M NaOH: 50 mg/mL

solubility

1 M HCl: 50 mg/mL

solubility

-


Still not finding the right product?

Explore all of our products under L-Aspartic acid


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
A7219-100G04061835568086
A7219-1KG04061833383827

Questions

  1. How to solubilize in water?

    1 answer
    1. L-Aspartic acid is soluble in water at 5 mg/mL.

      Helpful?

Reviews

No rating value

Active Filters