Skip to Content
Merck
All Photos(2)

Documents

A6150

Sigma-Aldrich

α1-Antitrypsin from human plasma

salt-free, lyophilized powder

Synonym(s):

α1-Proteinase inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

biological source

human plasma

Quality Level

form

salt-free, lyophilized powder

concentration

≥60% (biuret)

technique(s)

inhibition assay: suitable

UniProt accession no.

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Serine protease inhibitor; inhibits trypsin, chymotrypsin and pancreatic and granulocytic elastase, and acrosin. Effective concentration equimolar with proteinase.The effects of hereditary α1-antitrypsin deficiency and certain autoimmune states result from uncontrolled proteolysis in vivo. Direct α1-antitrypsin replacement therapy has shown promise in animal models of Type 1 diabetes.
≤10 mg will inhibit 1.0 mg of trypsin with activity of 10,000 BAEE units per mg protein. ≤10 mg will inhibit approx. 1.0 mg of ·α-chymotrypsin with activity of 40-50 BTEE units per mg protein.

Caution

Aqueous stock solutions containing 0.01% NaN3 are stable for several months. Solutions can be stored at −80 °C, but should not be refrozen. Unstable below pH 5.5. Inactivated by some non-serine proteinases and by oxidation of active site methionine residue.

Preparation Note

Chromatographically prepared and partially purified.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J M Bender et al.
Experimental mechanics, 61(1), 235-251 (2021-03-30)
Myxomatous mitral valve degeneration is a common cause of mitral regurgitation and is often associated with mitral valve prolapse. With no known targets to pharmacologically treat mitral valve prolapse, surgery is often the only treatment option. Recently, radiofrequency ablation has
Yukako Oda et al.
Science advances, 7(47), eabj6895-eabj6895 (2021-11-18)
[Figure: see text].
Kathirvel Alagesan et al.
Analytical and bioanalytical chemistry, 409(2), 529-538 (2016-12-03)
Glycopeptide enrichment is a crucial step in glycoproteomics for which hydrophilic interaction chromatography (HILIC) has extensively been applied due to its low bias towards different glycan types. A systematic evaluation of applicable HILIC mobile phases on glycopeptide enrichment efficiency and
Cynthia L Bristow et al.
Journal of immunology (Baltimore, Md. : 1950), 180(1), 492-499 (2007-12-22)
To identify positive or negative factors for HIV-1 infectivity, clones from the U937 promonocytic cell line that express similar levels of CD4 and CXCR4, but differ in HIV-1 susceptibility, were compared. In contrast to HIV-1 permissive clone 10 (plus), nonpermissive
Leslie W Chan et al.
Nature nanotechnology, 15(9), 792-800 (2020-07-22)
Human breath contains many volatile metabolites. However, few breath tests are currently used in the clinic to monitor disease due to bottlenecks in biomarker identification. Here we engineered breath biomarkers for respiratory disease by local delivery of protease-sensing nanoparticles to

Articles

Elastase application index for understanding leukocyte elastase, a 29KDa serine endoprotease.

Elastase application index for understanding leukocyte elastase, a 29KDa serine endoprotease.

Elastase application index for understanding leukocyte elastase, a 29KDa serine endoprotease.

Elastase application index for understanding leukocyte elastase, a 29KDa serine endoprotease.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service