Skip to Content
Merck

Skip To

A5757

Antifoam B Emulsion

aqueous-silicone emulsion

Synonym(s):

Antifoaming agent B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12161900
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

emulsion

contains

emulsifier (different from those present in Antifoam A emulsion)

technique(s)

cell culture | hybridoma: suitable, microbiological culture: suitable

viscosity

400 cP

density

1.01 g/mL

General description

Antifoam eliminates excessive foaming formed during impingement, due to proteins and/or carbohydrates in the collection medium. Antifoam B emulsion is a silicone based antifoam. Antifoam B emulsion does not affect the growth of bacteria. Antifoam B emulsion functions as an antifoaming agent in shake flask culture of bacteria.

Application

Antifoam B emulsion has been used:
  • to enhance collection efficiency in collection media
  • as a component in stress granule lysis buffer to reduce foaming, in host cell lysis buffer to reduce foaming caused by detergents
  • to improve solubility of antibiotics
  • as a foaming controls in bacterial growth media

Other Notes

A 10% aqueous emulsion of polydimethylsiloxane

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A8011A6582A8582
form

emulsion

form

suspension

form

emulsion (aqueous)

form

emulsion (aqueous)

technique(s)

cell culture | hybridoma: suitable, microbiological culture: suitable

technique(s)

cell culture | hybridoma: suitable, microbiological culture: suitable

technique(s)

cell culture | hybridoma: suitable, microbiological culture: suitable

technique(s)

cell culture | hybridoma: suitable

viscosity

400 cP

viscosity

400 cP

viscosity

-

viscosity

-

density

1.01 g/mL

density

1.01 g/mL

density

0.97 g/mL at 25 °C

density

1.0 g/mL at 25 °C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

contains

emulsifier (different from those present in Antifoam A emulsion)

contains

emulsifier

contains

-

contains

-


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

>214.0 °F - closed cup

flash_point_c

> 101.1 °C - closed cup

pictograms

Environment

signalword

Danger

Hazard Classifications

Aquatic Chronic 2 - PBT - vPvB



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Fermentation pH modulates the size distributions and functional properties of Gluconobacter albidus TMW 2.1191 levan
Ua-Arak T, et al.
Frontiers in Microbiology, 8, 807-807 (2017)
Isolation of yeast and mammalian stress granule cores
Wheeler JR, et al.
Methods, 126, 12-17 (2017)
Separation of metabolic supply and demand: aerobic glycolysis as a normal physiological response to fluctuating energetic demands in the membrane
Epstein T, et al.
Cancer & Metabolism, 2(1), 7-7 (2014)



Global Trade Item Number

SKUGTIN
A5757-250ML04061837596087
A5757-500ML04061837596094

Questions

1–3 of 3 Questions  
  1. Is the product autoclavable? Would it be autoclavable diluted in culture media, or is the product shipped sterile?

    1 answer
    1. This product is supplied non-sterile. However, it is autoclavable. Please note the emulsion may turn into gel at high heat but should return to normal fluidity once cooled.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters