Skip to Content
Merck

Skip To

A5472

Formalin Free Tissue Fixative

greener alternative
Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.47
UNSPSC Code:
12171500
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

colorless

density

1.044 g/cm3

application(s)

hematology
histology

greener alternative category

storage temp.

room temp

Application

A formalin-free tissue fixative that is a less toxic alternative to formalin. Performs well with PCR, in-situ hybridization and immunohistological staining.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of formalin for tissue processing.

Other Notes

Alcoholic solution with hydroxylated compounds. The product is ready to use and requires no dilution or preparation.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HT5012HT501320Z2902
color

colorless

color

colorless

color

colorless

color

-

form

solution

form

-

form

-

form

-

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

application(s)

hematology
histology

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

storage temp.

room temp

density

1.044 g/cm3

density

1.080 g/cm3

density

1.080 g/cm3

density

-

sustainability

Greener Alternative Product

sustainability

-

sustainability

-

sustainability

-


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 3

flash_point_f

118.0 °F

flash_point_c

47.8 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters