Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Level
assay
≥97% (HPLC)
form
powder
composition
Protein Content, ≥80%
UniProt accession no.
storage temp.
−20°C
SMILES string
CCC(C)C(NC(=O)C(NC(=O)C(C)NC(=O)CNC(=O)C(CCCCN)NC(=O)C(CC(N)=O)NC(=O)C(CO)NC(=O)CN)C(C)CC)C(=O)NCC(=O)NC(CC(C)C)C(=O)NC(CCSC)C(O)=O
InChI
1S/C45H81N13O14S/c1-9-24(5)36(43(69)50-21-35(63)52-29(17-23(3)4)40(66)55-28(45(71)72)14-16-73-8)58-44(70)37(25(6)10-2)57-38(64)26(7)51-34(62)20-49-39(65)27(13-11-12-15-46)54-41(67)30(18-32(48)60)56-42(68)31(22-59)53-33(61)19-47/h23-31,36-37,59H,9-22,46-47H2,1-8H3,(H2,48,60)(H,49,65)(H,50,69)(H,51,62)(H,52,63)(H,53,61)(H,54,67)(H,55,66)(H,56,68)(H,57,64)(H,58,70)(H,71,72)
InChI key
WIHBNMPFWRHGDF-UHFFFAOYSA-N
Gene Information
human ... APP(351)
General description
Application
- to induce neurotoxicity in cortical cultures
- to induce Alzheimer′s disease in rat model
- to induce apoptosis in mesenchymal stem cells (MSCs)
Biochem/physiol Actions
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥97% (HPLC) | assay ≥95% (HPLC) | assay ≥90% (HPLC) | assay ≥95% (HPLC) |
| form powder | form powder | form lyophilized powder | form powder |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| Gene Information human ... APP(351) | Gene Information human ... APP(351) | Gene Information human ... APP(351) | Gene Information human ... APP(351) |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| A4559-250UG | 04061833374665 |
| A4559-1MG | 04061838443540 |



