Skip to Content
Merck

A4544

L-Ascorbic acid

suitable for cell culture, suitable for plant cell culture, ≥98%

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
UNSPSC Code:
12352201
NACRES:
NA.21
PubChem Substance ID:
EC Number:
200-066-2
Beilstein/REAXYS Number:
84272
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98%

form

powder

mol wt

Mw 176.12 g/mol

technique(s)

cell culture | mammalian: suitable, cell culture | plant: suitable

color

white to light yellow

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

water: soluble 50 mg/mL

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

General description

L-ascorbic acid also called vitamin C, is a water soluble vitamin and acts as a reducing agent for various biochemical reactions. Ascorbic acid plays a key role in the production of collagen. Ascorbic acid is essential for neurodevelopment, neurotransmitter synthesis and modulates neuronal synapsis. Dietary inadequacy of vitamin C has negative impact on cognition.

Application

L-ascorbic acid has been used as an ingredient in the basal medium for bovine mammary epithelial cells and oocyte-granulosa cell complexes. It has been used as a supplement in the osteogenic medium for osteogenic differentiation.

Biochem/physiol Actions

Ascorbic Acid, also known as Vitamin C, is a six-carbon lactone produced by plants and some animal species but not by humans and other primates. Ascorbic acid functions as an enzymatic cofactor for multiple enzymes, serving as an electron donor for monooxygenases and dioxygenases. Ascorbic acid also functions as a powerful antioxidant, particularly in regards to reactive oxygen species.


Still not finding the right product?

Explore all of our products under L-Ascorbic acid


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
A4544-25G04061835542130
A4544-500G04061835542147
A4544-100G04061835542123