Skip to Content
Merck

Skip To

A3543

Anti-Human IgG (whole molecule)−Agarose antibody produced in goat

IgG fraction of antiserum, PBS suspension

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
agarose conjugate
Clone:
polyclonal
Application:
IEP
Citations:
8
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

agarose conjugate

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

PBS suspension

species reactivity

human

extent of labeling

~2.4 mg Ab per mL

technique(s)

immunoelectrophoresis: suitable

capacity

1 mg/mL binding capacity

storage temp.

2-8°C

target post-translational modification

unmodified

General description

IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections.

Immunogen

Pooled normal human serum

Application

Anti-human IgG (whole molecule) is suitable for immunoelectrophoresis. Antibody depletion from normal human serum was carried out by treating diluted serum with 50 microl of agarose-linked goat anti-human IgG (A3543) for 90 minutes at 4 °.
Antibody depletion from normal human serum was carried out by treating diluted serum with 50 μl of agarose-linked goat anti-human IgG (A3543) for 90 minutes at 4 degrees.

Biochem/physiol Actions

Anti-human IgG recognizes human IgG (whole molecules). Goat anti-human is purified by ion exchange chromatography and then covalently bound to agarose.
IgG stimulates the classical pathway of the complement system. It neutralizes virus particles and toxins. IgG plays a crucial role in antibody-dependent cell-mediated cytotoxicity (ADCC). IgG might be associated with allergy. It has the longest serum half life.. Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections.

Physical form

Suspension in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
I3382A8914A9294
species reactivity

human

species reactivity

-

species reactivity

-

species reactivity

goat

biological source

goat

biological source

goat

biological source

goat

biological source

rabbit

technique(s)

immunoelectrophoresis: suitable

technique(s)

Ouchterlony double diffusion: suitable

technique(s)

immunoelectrophoresis: suitable

technique(s)

immunoelectrophoresis: suitable

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

conjugate

agarose conjugate

conjugate

unconjugated

conjugate

agarose conjugate

conjugate

agarose conjugate

form

PBS suspension

form

lyophilized powder

form

saline suspension

form

PBS suspension


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
A3543-10ML04061833361047

Questions

Reviews

No rating value

Active Filters