Skip to Content
Merck

Skip To

A2638

N-Acetyl-DL-serine

Synonym(s):

N-acetyl-serine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H9NO4
CAS Number:
Molecular Weight:
147.13
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352209
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥99% (TLC)

Quality Segment

form

powder

technique(s)

LC/MS: suitable

color

white

application(s)

cell analysis

storage temp.

−20°C

SMILES string

CC(=O)NC(CO)C(O)=O

InChI

1S/C5H9NO4/c1-3(8)6-4(2-7)5(9)10/h4,7H,2H2,1H3,(H,6,8)(H,9,10)

InChI key

JJIHLJJYMXLCOY-UHFFFAOYSA-N

Biochem/physiol Actions

N-Acetyl-DL-serine is a racemic mixture of N-acetylated D and L serine that may be used in nutrition, physicochemical and racemic separation research.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A6262A4021A2010
form

powder

form

powder

form

powder

form

powder

assay

≥99% (TLC)

assay

≥98.0% (TLC)

assay

≥98% (TLC)

assay

≥98% (TLC)

technique(s)

LC/MS: suitable

technique(s)

ligand binding assay: suitable

technique(s)

-

technique(s)

ligand binding assay: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

color

white

color

white to off-white

color

colorless to white

color

colorless to white

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters