Skip to Content
Merck

Skip To

A2429

Anti-Mouse IgG (Fc specific)–Alkaline Phosphatase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgG (Fc specific)–AP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
MDL number:
Conjugate:
alkaline phosphatase conjugate
Clone:
polyclonal
Application:
ELISA (d), IHC (p), WB CL
Citations:
45
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

goat

Quality Segment

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

should not react with

bovine, human, horse

technique(s)

direct ELISA: 1:30,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:40, western blot (chemiluminescent): 1:160,000-1:320,000

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It is a glycoprotein. IgG is a major class of immunoglobulin. Mouse consists of five immunoglobulin classes- IgM, IgG, IgA, IgD and IgE. Mouse IgG is further divided into five classes- IgG1, IgG2a, IgG2b and IgG3.
Immunoglobulin G (IgG) is a glycoprotein antibody that regulates immune responses such as phagocytosis and is also involved in the development of autoimmune diseases. IgG1 regulates complement fixation in mice.
Mouse IgG is a plasma B cell derived antibody isotype defined by its heavy chain. IgG is the most abundant antibody isotype found in mouse serum. IgG crosses the placental barrier, is a complement activator and binds to the Fc-receptors on phagocytic cells. The level of IgG may vary with the status of disease or infection.

Immunogen

Purified mouse IgG, Fc fragment

Application

Alkaline phosphatase-conjugated goat anti-mouse Fc specific antibody was used as a secondary antibody in ELISA assays at a dilution of 1:1000 in PBS/0.1% Tween and 1% BSA for 1.5 hours at 37°C. Antibody was developed using 4-nitrophenyl phosphate (Sigma) as a substrate for 30 minutes at 37°C.
Anti-Mouse IgG (Fc specific)-Alkaline Phosphatase antibody is suitable for use in ELISA (1:1500) and western blot. The antibody can also be used for immunohistochemistry (1:40 using formalin-fixed, paraffin-embedded sections).
Anti-Mouse IgG (Fc specific)-Alkaline Phosphatase antibody produced in goat has been used in enzyme-linked immunosorbent assay (ELISA), sandwich ELISA and enzyme-linked immune absorbent spot (ELISPOT).
The product has been used as a secondary antibody for detecting mouse serum anti-hemagglutinin (HA) antibodies using ELISA techniques.

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III reactions. IgG helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Physical form

Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with bovine, horse, or human samples.

Other Notes

Antibody adsorbed with bovine, equine and human serum proteins.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A7434A1418A1682
biological source

goat

biological source

goat

biological source

goat

biological source

goat

technique(s)

direct ELISA: 1:30,000, western blot (chemiluminescent): 1:160,000-1:320,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:40

technique(s)

direct ELISA: 1:40,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:20, western blot (chemiluminescent): 1:30,000 using β-actin in total cell extract of HiLa cells (5-10 μg per well)

technique(s)

direct ELISA: 1:40,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25, western blot: 1:50,000-1:100,000 using total cell extract of HeLa cells

technique(s)

direct ELISA: 1:40,000, immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:10, western blot: 1:200,000-1:400.000 using assay detecting β-Actin in total cell extract of HeLa cells (5-10 μg per well)

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

alkaline phosphatase conjugate

conjugate

alkaline phosphatase conjugate

conjugate

alkaline phosphatase conjugate

conjugate

alkaline phosphatase conjugate

species reactivity

mouse

species reactivity

-

species reactivity

mouse

species reactivity

mouse

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

Instructions






Global Trade Item Number

SKUGTIN
A2429-.25ML04061838352156
A2429-1ML04061833350300
A2429-.5ML04061833350294

Questions

Reviews

No rating value

Active Filters