Skip to Content
Merck

Skip To

A2033

Alginic acid sodium salt from brown algae

Medium viscosity

Synonym(s):

Algin, Sodium alginate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

algae (brown)

Quality Segment

form

powder

color

white to brown

viscosity

≥2,000 cP, 2 %(25 °C)

solubility

water: 10 mg/mL, slightly hazy to strongly hazy, faintly yellow to yellow

application(s)

advanced drug delivery

storage temp.

2-8°C

SMILES string

[Na+].[O-]C(=O)C1O[C@H]([C@H]([C@H]([C@@H]1O)O)O)O

InChI

1S/C6H10O7.Na/c7-1-2(8)4(5(10)11)13-6(12)3(1)9;/h1-4,6-9,12H,(H,10,11);/q;+1/p-1/t1-,2-,3-,4?,6+;/m0./s1

InChI key

MSXHSNHNTORCAW-MPGIDXPLSA-M

Application

Alginate, a colloidal polyuronic acid structural molecule capable of gelation, is used in the preparation of colloidal biodegradable structures such as gels, biofilms, beads, nanoparticles, and microcapsules suitable for applications that range from gel based separation technologies to drug delivery and cell preservation.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.
A straight-chain, hydrophilic, colloidal, polyuronic acid composed of guluronic and mannuronic acid residues.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A111271238180947
biological source

algae (brown)

biological source

algae (brown)

biological source

algae (brown)

biological source

algae (marine)

solubility

water: 10 mg/mL, slightly hazy to strongly hazy, faintly yellow to yellow

solubility

water: 10 mg/mL, cloudy to hazy, faintly yellow

solubility

-

solubility

-

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

form

powder

form

powder

form

powder

form

powder

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

storage temp.

-

color

white to brown

color

white to brown

color

white to light beige

color

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 14 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. iam master student , want to study drag reduction in water pipeline by using bio-polymer , i want Sodium Alginate like the above but: with high molecule weight ( million Da) , and tell me if its rigid or flexible( M/G=?) , and must be pure.. thank you

    1 answer
    1. The molecular weight and M/G ratio are not tested for this sodium alginate. Generally, the molecular weight for sodium alginate with a viscosity over 2000 (2% solution) is over 50,000 Da, and the M/G ratio is about 65:35. Most alginic acid materials have a molecular weight of approximately 100 - 200 kDa, but this range is not guaranteed.

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. what is the viscosity?

    1 answer
    1. As listed in the 'PROPERTIES' section, the viscosity is ?2,000 cP. The exact value is listed on the lot specific Certificate of Analysis. Please navigate to the DOCUMENTATION section of the Product Detail Page to access a Certificate under ‘Certificate of Analysis’: https://www.sigmaaldrich.com/product/sigma/a2033#product-documentation

      Helpful?

  5. A2033 제품의 밀도를 알고 싶습니다 정확한 값까지는 아니어도 추정치 만이라도 알고 싶습니다

    1 answer
    1. Unfortunately, the density of this product is not determined internally. However, various external sources state that Sodium Alginate has a density of 1.0 g/cm3(Temp: 25 °C).

      Helpful?

  6. What is the average molecular weight of this product?

    1 answer
    1. The Molecular Weight of this product is approximately 200,000 daltons.

      Helpful?

  7. What is the purity and the loss on drying of the product A2033? Additionally, what is the ash content?

    1 answer
    1. The purity and loss on drying are not tested for alginic acid. Please see the link below to review the testing and specifications for this product: https://www.sigmaaldrich.com/specification-sheets/235/361/A2033-BULK________SIGMA____.pdf

      Helpful?

  8. How to sterilization this product without properties interference?

    1 answer
    1. Solutions may be filter-sterilized or autoclaved prior to the addition of cations (required for gelation). Note that extended autoclave times may be detrimental to bead formation.

      Helpful?

  9. Does this product create a low or high stiffness gel?

    1 answer
    1. This product is not tested for gelation or gel strength. Alginate forms a gel in the presence of cations. Typically CaCl2 is used to initiate gel formation. Increasing the concentration of calcium will result in a more firm gel. Calcium chloride may be added at a range of 0.1M to 0.5M, with gel strength increasing with the CaCl2 concentration.

      Helpful?

  10. What is the ratio of mannuronic acid to guluronic acid in this product (A2033)?

    1 answer
    1. The A2033 has a mannuronic acid to guluronic acid ratio of approximately 1.56. The mannuronic acid range is 60-70 % and guluronic acid range is 30-40 %.

      Helpful?

1–10 of 14 Questions  

Reviews

No rating value

Active Filters