A1879
L-2-Aminobutyric acid
≥99% (titration), suitable for ligand binding assays
Synonym(s):
L-α-Aminobutyric acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H9NO2
CAS Number:
Molecular Weight:
103.12
Beilstein:
1720935
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
L-2-Aminobutyric acid, ≥99% (titration)
Assay
≥99% (titration)
form
powder
technique(s)
ligand binding assay: suitable
color
white
application(s)
peptide synthesis
SMILES string
CC[C@H](N)C(O)=O
InChI
1S/C4H9NO2/c1-2-3(5)4(6)7/h3H,2,5H2,1H3,(H,6,7)/t3-/m0/s1
InChI key
QWCKQJZIFLGMSD-VKHMYHEASA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Targeted metabolomics combined with machine learning to identify and validate new biomarkers for early SLE diagnosis and disease activity.: This study utilizes L-2-aminobutyric acid in targeted metabolomics, enhancing the predictive accuracy for Systemic Lupus Erythematosus (SLE), a crucial development in autoimmune disease diagnostics (Liang J et al., 2024).
- Alcoholamine enhanced fractionation of cellulose from lignocellulosic biomass in ionic liquids.: This research highlights the use of L-2-aminobutyric acid in the process of biomass conversion, offering insights into more efficient biofuel production techniques (Zhu Y et al., 2023).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Carl Henrik Görbitz
Acta crystallographica. Section B, Structural science, 66(Pt 2), 253-259 (2010-03-23)
The crystal structure of L-2-aminobutyric acid, an L-alanine analogue with an ethyl rather than a methyl side chain, has proved elusive owing to problems growing diffraction quality crystals. Good diffraction data have now been obtained for two polymorphs, in space
Danica P Galonić et al.
Nature chemical biology, 3(2), 113-116 (2007-01-16)
Enzymatic incorporation of a halogen atom is a common feature in the biosyntheses of more than 4,500 natural products. Halogenation of unactivated carbon centers in the biosyntheses of several compounds of nonribosomal peptide origin is carried out by a class
Izumi Kawabata et al.
Nature communications, 3, 722-722 (2012-03-08)
Synaptic remodelling coordinated with dendritic growth is essential for proper development of neural connections. After establishment of synaptic contacts, synaptic junctions are thought to become stationary and provide fixed anchoring points for further dendritic growth. However, the possibility of active
Cornelia Reimmann et al.
Journal of bacteriology, 186(19), 6367-6373 (2004-09-18)
In Pseudomonas aeruginosa, the antibiotic dihydroaeruginoate (Dha) and the siderophore pyochelin are produced from salicylate and cysteine by a thiotemplate mechanism involving the peptide synthetases PchE and PchF. A thioesterase encoded by the pchC gene was found to be necessary
Francesco Gasparrini et al.
Journal of the American Chemical Society, 130(2), 522-534 (2007-12-22)
The structure, stability, and reactivity of proton-bound diastereomeric [M x H x A]+ complexes between some amino acid derivatives (A) and several chiral tetra-amide macrocycles (M) have been investigated in the gas phase by ESI-FT-ICR and ESI-ITMS-CID mass spectrometry. The
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service