Skip to Content
Merck

Skip To

97318

D-Mannoheptulose

≥99.0% (HPLC)

Synonym(s):

D-Manno-2-heptulose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C7H14O7
CAS Number:
Molecular Weight:
210.18
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
EC Number:
222-795-5
Beilstein/REAXYS Number:
1726433
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥99.0% (HPLC)

form

crystals

color

white

mp

151  °C ((304 °F ))

storage temp.

room temp

SMILES string

OC[C@H]1OC(O)(CO)[C@@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C7H14O7/c8-1-3-4(10)5(11)6(12)7(13,2-9)14-3/h3-6,8-13H,1-2H2/t3-,4-,5+,6+,7?/m1/s1

InChI key

HAIWUXASLYEWLM-BWSJPXOBSA-N

Application

D-Mannoheptulose, a heptose, may be used as a hexokinase inhibitor (like glucosamine) in studies involving glucose response, glucose metabolism, glucose signaling and glucose sensing mechanisms. Analogs of D-mannoheptulose may be developed for use in noninvasive imaging of pancreatic beta cells.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
635798626578832
assay

≥99.0% (HPLC)

assay

≥99.5% (sum of enantiomers, HPLC)

assay

≥99.0% (HPLC)

assay

≥90% (TLC)

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

100

form

crystals

form

solid

form

crystals

form

powder

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

−20°C

color

white

color

white

color

white

color

white to light yellow

mp

151  °C ((304 °F ))

mp

133-140 °C (lit.)

mp

124-127 °C

mp

-


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
97318-25MG04061833324967

Questions

  1. What's the solubility of the product? what solvent is convenient touse?

    1 answer
    1. The product is not tested for solubility. However, according to a literature search, this material is soluble in DMF at 20 mg/ml, in DMSO at 20 mg/ml, in Ethanol at 1 mg/ml, and in PBS, pH 7.2, at 10 mg/ml. It would be up to the end user to determine the most appropriate solvent for a specific application.

      Helpful?

Reviews

No rating value

Active Filters