Skip to Content
Merck

Skip To

94046

Sodium phosphate dibasic solution

BioUltra, 0.5 M in H2O

Synonym(s):

Disodium hydrogen phosphate solution, di-Sodium hydrogen phosphate solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
UNSPSC Code:
12161700
eCl@ss:
38070211
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioUltra

Quality Segment

form

liquid

concentration

0.5 M in H2O

impurities

insoluble matter, passes filter test, ≤0.001% total nitrogen (N)

pH

8.8-9.4 (25 °C, 0.5 M in H2O)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

density

1.064 g/mL at 20 °C

anion traces

chloride (Cl-): ≤10 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg, As: ≤0.1 mg/kg, Ba: ≤1 mg/kg, Bi: ≤1 mg/kg, Ca: ≤5 mg/kg, Cd: ≤1 mg/kg, Co: ≤1 mg/kg, Cr: ≤1 mg/kg, Cu: ≤1 mg/kg, Fe: ≤1 mg/kg, K: ≤20 mg/kg, Li: ≤1 mg/kg, Mg: ≤1 mg/kg, Mn: ≤1 mg/kg, Mo: ≤1 mg/kg, Ni: ≤1 mg/kg, Sr: ≤1 mg/kg, Zn: ≤1 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: 0.019, λ: 280 nm Amax: 0.016

application(s)

diagnostic assay manufacturing

SMILES string

[Na+].[Na+].OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Application

Sodium phosphate dibasic solution has been used:
  • to supplement Dulbecco′s Modified Eagle Medium (DMEM) in calcifying medium to induce the calcification of vascular smooth muscle cells (VSMCs)
  • to prepare a stock solution of CaCl2 and MgCl2 to synthesize calcium phosphate (CPB) and magnesium phosphate bions (MPBs) respectively
  • to supplement 2xYT agar plates to culture BL21 DE3 star cells and BL21dLac Gold cells

Sodium phosphate dibasic solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S3264S5136RDD038
concentration

0.5 M in H2O

concentration

-

concentration

-

concentration

-

product line

BioUltra

product line

-

product line

BioReagent

product line

BioReagent, Redi-Dri

Quality Level

100

Quality Level

300

Quality Level

200

Quality Level

100

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

form

liquid

form

powder

form

powder or crystals

form

powder or crystals

density

1.064 g/mL at 20 °C

density

-

density

-

density

-


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
94046-100ML-F04061838170309
94046-500ML-F04061833283745

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters