Skip to Content
Merck

Skip To

93482

Phenylmethanesulfonyl fluoride solution

~0.1 M in ethanol (T)

Synonym(s):

Benzylsulfonyl fluoride, PMSF

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.77
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2088311
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

biological source

microbial, synthetic

form

liquid

concentration

~0.1 M in ethanol (T)

density

0.797 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

FS(=O)(=O)Cc1ccccc1

InChI

1S/C7H7FO2S/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

YBYRMVIVWMBXKQ-UHFFFAOYSA-N

Application

Phenylmethanesulfonyl fluoride solution has been used as a component in the cell lysate buffer in breast adenocarcinoma cell line, fibrosarcoma cell line and D5 hindbrain and spinal cord neural progenitors.

Biochem/physiol Actions

Administration of PMSF produces analgesia unrelated to its anticholinesterase effect, and prolongs the analagesic effect of centrally administered β-endorphin.[1]
Phenylmethanesulfonyl fluoride (PMSF) is a serine protease inhibitor. It binds covalently to active site residue of proteases and prevents proteolytic activity. PMSF is used in cell lysate buffers during protein purification to prevent target protein degradation.[2] PMSF effectively inhibits the naloxone-precipitated withdrawal jumping.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7883077004P7626
biological source

microbial, synthetic

biological source

synthetic

biological source

-

biological source

synthetic

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

300

form

liquid

form

powder or crystals

form

solution

form

powder

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

storage temp.

-

density

0.797 g/mL at 20 °C

density

-

density

-

density

-

concentration

~0.1 M in ethanol (T)

concentration

-

concentration

~50% in acetonitrile

concentration

-


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

53.6 °F - closed cup

flash_point_c

12 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



DNA-encoded library-derived DDR1 inhibitor prevents fibrosis and renal function loss in a genetic mouse model of Alport syndrome
Richter H, et al.
ACS Chemical Biology (2018)
Nervous system regionalization entails axial allocation before neural differentiation
Metzis V, et al.
Cell, 175(4), 1105-1118 (2018)
SREBP1 drives KRT80-dependent cytoskeletal changes and invasive behavior in endocrine resistant ERalpha breast cancer
Magnani L, et al.
bioRxiv, 380634-380634 (2018)



Global Trade Item Number

SKUGTIN
93482-250ML-F04061833282823
93482-50ML-F04061833282830

Questions

1–4 of 4 Questions  
  1. Is this product 93482 RNase-free?

    1 answer
    1. This product is not tested for RNase activity. Currently, an RNAse tested PMSF product is not available.

      Helpful?

  2. What can Product 93482, Phenylmethanesulfonyl fluoride solution, be used for?

    1 answer
    1. PMSF is a serine protease inhibitor, inhibiting the activity of enzymes such as thrombin, trypsin, papain, chymotrypsin and proteinase K. It also inhibits mammalian acetylcholinesterases.

      Helpful?

  3. How can I further dilute Product 93482, Phenylmethanesulfonyl fluoride solution?

    1 answer
    1. We would recommend that you use an anhydrous ethanol to further dilute this solution, since PMSF is very unstable in water.

      Helpful?

  4. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters