Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.47
UNSPSC Code:
12352106
Pricing and availability is not currently available.
Quality Level
shelf life
Expiry date on the label.
IVD
for in vitro diagnostic use
dilution
(for histology)
application(s)
hematology
histology
shipped in
wet ice
storage temp.
2-8°C
Application
Leukocyte Alkaline Phosphatase (LAP) kits are intended for the qualitative demonstration of alkaline phosphatase activity in white blood cells.
Peripheral blood or bone marrow preparations are fixed to a microscope slide. The film is then incubated in a mixture of naphthol AS-MX phosphate alkaline solution with fast blue RR salt. The resulting blue, insoluble, granular dye deposit indicates sites of alkaline phosphatase activity.
1 of 1
This Item | |||
|---|---|---|---|
| application(s) hematology | application(s) hematology | application(s) hematology | application(s) hematology |
| Quality Level 500 | Quality Level 500 | Quality Level 500 | Quality Level 500, 600 |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| IVD for in vitro diagnostic use | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use | IVD for in vitro diagnostic use |
| shelf life Expiry date on the label. | shelf life Expiry date on the label. | shelf life Expiry date on the label. | shelf life Expiry date on the label. |
Still not finding the right product?
Explore all of our products under Leukocyte Alkaline Phosphatase Kit
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lee W Tien et al.
Macromolecular bioscience, 12(12), 1671-1679 (2012-10-17)
The alignment and osteogenic differentiation of MSCs on patterned silk films (PF) is investigated as a bottom-up approach toward engineering bone lamellae. Screening PF with various groove dimensions shows that cell alignment is mediated by both the pattern width and
Alban J Smith et al.
Scientific reports, 7, 41872-41872 (2017-02-06)
The ability to isolate specific, viable cell populations from mixed ensembles with minimal manipulation and within intra-operative time would provide significant advantages for autologous, cell-based therapies in regenerative medicine. Current cell-enrichment technologies are either slow, lack specificity and/or require labelling.
Clarissa S Craft et al.
The Journal of biological chemistry, 285(31), 23858-23867 (2010-05-27)
MAGP1 is an extracellular matrix protein that, in vertebrates, is a ubiquitous component of fibrillin-rich microfibrils. We previously reported that aged MAGP1-deficient mice (MAGP1Delta) develop lesions that are the consequence of spontaneous bone fracture. We now present a more defined
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 85L1-1KT | 04061833032565 |




