Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Segment
assay
≥99.0% (HPLC)
form
solid
mol wt
134.13 g/mol
concentration
≤100% (o-phthaldialdehyde)
impurities
≤0.5% phthalic acid
color
white to light yellow
bp
83 °C/1.067 hPa
mp
54-56 °C
solubility
ethanol: 0.5 g/10 mL, clear, colorless to greenish-yellow, water: 37.7 g/L at 20 °C (OECD Test Guideline 105)
density
1.13 g/cm3 at 20 °C
fluorescence
λex 334 nm; λem 455 nm (Thiol Adduct), λex 340 nm; λem 450 nm in reaction buffer (with glycine)
suitability
suitable for fluorescence
storage temp.
2-8°C
SMILES string
O=Cc1ccccc1C=O
InChI
1S/C8H6O2/c9-5-7-3-1-2-4-8(7)6-10/h1-6H
InChI key
ZWLUXSQADUDCSB-UHFFFAOYSA-N
General description
Application
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥99.0% (HPLC) | assay - | assay ≥97.5% (GC) | assay - |
| solubility ethanol: 0.5 g/10 mL, clear, colorless to greenish-yellow, water: 37.7 g/L at 20 °C (OECD Test Guideline 105) | solubility 53 g/L | solubility 53 g/L | solubility - |
| Quality Level 200 | Quality Level 100 | Quality Level 200 | Quality Level 200 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-30°C | storage temp. 2-8°C |
| bp 83 °C/1.067 hPa | bp 83-84 °C/1 hPa | bp 83-84 °C/1 hPa | bp - |
| form solid | form solid | form solid | form - |
Still not finding the right product?
Explore all of our products under Phthaldialdehyde
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk
WGK 3
flash_point_f
269.6 °F - closed cup
flash_point_c
132 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 79760-5G | 04061835570799 |
| 79760-1G | 04061832964812 |
| 79760-50G | 04061835183937 |




