Skip to Content
Merck

Skip To

75742

Esterase Pseudomonas fluorescens, recombinant from E. coli

≥4 U/mg

Synonym(s):

Carboxylic ester hydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-773-7
MDL number:
EC Number:
Specific activity:
≥4 U/mg
Recombinant:
expressed in E. coli
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

Quality Level

form

powder

specific activity

≥4 U/mg

storage temp.

2-8°C

Application

The compound is commonly used for the synthesis of biodiesel and biopolymers, as well as in the production of pharmaceuticals, agrochemicals and flavor compounds.

Biochem/physiol Actions

Esterase acts on water-soluble carboxyl esters containing short chain fatty acids.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

1 U corresponds to the amount of enzyme which liberates 1 μmol acetic acid from p-nitrophenylacetate per minute at pH 7.5 and 25°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4605143388
specific activity

≥4 U/mg

specific activity

≥0.2 U/mg

specific activity

≥30.0 U/g

form

powder

form

lyophilized

form

crystalline, crystals, powder or flakes

recombinant

expressed in E. coli

recombinant

-

recombinant

expressed in E. coli

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

Quality Level

100

Quality Level

-

Quality Level

100


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Marie C Fortin et al.
Drug metabolism and disposition: the biological fate of chemicals, 41(2), 326-331 (2012-12-12)
Studies on therapeutic drug disposition in humans have shown significant alterations as the result of pregnancy. However, it is not known whether pesticide metabolic capacity changes throughout pregnancy, which could affect exposure of the developing brain. We sought to determine
B Sànchez-Nogué et al.
Environmental science and pollution research international, 20(5), 3480-3488 (2012-12-06)
The common sole, Solea solea (Linneus, 1758), and the Senegalese sole, Solea senegalensis (Kaup, 1858), are two important commercial species that coexist in the NW Mediterranean. In order to assess the species' ability to respond to chemical insults, a comparison
Judit Marsillach et al.
Toxicology, 307, 46-54 (2012-12-25)
Exposure to organophosphorus (OP) compounds can lead to serious neurological damage or death. Following bioactivation by the liver cytochromes P450, the OP metabolites produced are potent inhibitors of serine active-site enzymes including esterases, proteases and lipases. OPs may form adducts



Global Trade Item Number

SKUGTIN
75742-500MG04061833217283
75742-10MG-KC04061824212914
75742-100MG04061832916576

Questions

Reviews

No rating value

Active Filters