Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
Vibrio cholerae
form
liquid
specific activity
≥1.5 U/mg protein
concentration
≥1.5 U/mL
density
1.00 g/mL at 20 °C
storage temp.
2-8°C
Application
1 of 1
This Item | |||
|---|---|---|---|
| specific activity ≥1.5 U/mg protein | specific activity 2-10 units/mg protein (mucin), 6-15 units/mg protein (using 4MU-NANA) | specific activity 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin) | specific activity ≥0.1 units/mg solid (using mucin), ≥1.3 units/mg solid (using 4MU-NANA) |
| biological source Vibrio cholerae | biological source Clostridium perfringens str. 13 | biological source - | biological source - |
| concentration ≥1.5 U/mL | concentration - | concentration - | concentration - |
| form liquid | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| density 1.00 g/mL at 20 °C | density - | density - | density - |
Still not finding the right product?
Explore all of our products under Neuraminidase from Vibrio cholerae
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

