Skip to Content
Merck

Skip To

72068

Sodium hydroxide solution

BioUltra, Molecular Biology, 10 M in H2O

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00
NACRES:
NA.26
PubChem Substance ID:
UNSPSC Code:
12352316
MDL number:
Concentration:
10 M in H2O, 9500-10500 mmol/L (titration, acidimetric)
Form:
liquid
Grade:
Molecular Biology
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

Molecular Biology

Quality Segment

vapor pressure

3 mmHg ( 37 °C)

product line

BioUltra

form

liquid

concentration

10 M in H2O, 9500-10500 mmol/L (titration, acidimetric)

technique(s)

ELISA: suitable, immunofluorescence: suitable, western blot: suitable

impurities

DNases, none detected, RNases, none detected, insoluble matter, passes filter test, phosphatases, none detected, proteases, none detected

pH

14

bp

105-140 °C

mp

-12 - -10 °C (10 - 50 °F)

solubility

water: soluble

density

1.320-1.340 g/cm3

cation traces

Ba: ≤1 mg/kg, Bi: ≤1 mg/kg, Ca: ≤5 mg/kg, Cd: ≤1 mg/kg, Co: ≤1 mg/kg, Cr: ≤1 mg/kg, Cu: ≤1 mg/kg, Fe: ≤1 mg/kg, K: ≤100 mg/kg, Li: ≤1 mg/kg, Mg: ≤1 mg/kg, Mn: ≤1 mg/kg, Mo: ≤1 mg/kg, Ni: ≤1 mg/kg, Pb: ≤1 mg/kg, Sr: ≤1 mg/kg, Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: ≤0.08, λ: 280 nm Amax: ≤0.05

SMILES string

[OH-].[Na+]

suitability

suitable for Western blot, suitable for molecular biology

application(s)

advanced drug delivery
agriculture
life science and biopharma

InChI

1S/Na.H2O/h;1H2/q+1;/p-1

InChI key

HEMHJVSKTPXQMS-UHFFFAOYSA-M

General description

Sodium hydroxide (NaOH) is an inorganic, white solid compound, which is found as pellets, flakes, and granules. This highly caustic metallic base and alkali can readily absorb moisture and carbon dioxide in the air. NaOH is deliquescent and soluble in ethanol, water, and methanol.

Application

Sodium hydroxide solution has been used:
  • to prepare reagent A (0.1 M NaOH) for the pH inactivation of endogenous L-lactate by L-lactate dehydrogenase (L-LDH)[1]
  • as a component of fresh alkaline melting solution for the denaturation of the antisense strand[2]
  • to prepare Type-I collagen hydrogels for microfluidic device fabrication[3]

Biochem/physiol Actions

Sodium hydroxide (NaOH) is used in various industries, including textiles, drinking water, soaps, and detergents, pulp and paper manufacturing, and as a drain cleaner.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S27707206479724
grade

Molecular Biology

grade

-

grade

-

grade

-

concentration

10 M in H2O, 9500-10500 mmol/L (titration, acidimetric)

concentration

1.0 N

concentration

49-51% in water

concentration

0.9-1.1 M

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

form

liquid

form

solution

form

liquid

form

liquid

solubility

water: soluble

solubility

-

solubility

water: soluble (completely miscible)

solubility

-

pH

14

pH

-

pH

14.0

pH

-


Still not finding the right product?

Explore all of our products under Sodium hydroxide solution


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Rikke Wehner Rasmussen et al.
Scandinavian journal of clinical and laboratory investigation, 81(4), 312-317 (2021-04-22)
Intestinal infarction is the fast-evolving endpoint of impaired blood perfusion to an intestinal segment which may have fatal outcome. Early diagnosis and treatment within 6 h reduce mortality. Currently, d-lactate is a promising biomarker, however, not available in the acute clinical
Possible materials for producing Geopolymer concrete and its performance with and without Fibre addition-A State of the art review
Kirupa JP Alwis Deva and Sakthieswaran N
International journal of civil and structural engineering, 5(3), 296-296 (2015)
Xin Cui et al.
Biomaterials, 161, 164-178 (2018-02-09)
Glioblastoma (GBM) is the most lethal primary adult brain tumor and its pathology is hallmarked by distorted neovascularization, diffuse tumor-associated macrophage infiltration, and potent immunosuppression. Reconstituting organotypic tumor angiogenesis models with biomimetic cell heterogeneity and interactions, pro-/anti-inflammatory milieu and extracellular



Global Trade Item Number

SKUGTIN
72068-100ML04061838254276

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. where is user manual?

    1 answer
    1. This product does not have a protocol associated with it. You may find it helpful to view the peer reviewed articles that mention the use of this product: https://www.sigmaaldrich.com/search/72068?focus=papers&page=1&perpage=30&sort=relevance&term=72068&type=citation_search.

      Helpful?

Reviews

No rating value

Active Filters