Skip to Content
Merck

Skip To

71859

Sodium (meta)periodate

BioUltra, ≥99.5% (RT)

Synonym(s):

Sodium periodate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
NaIO4
CAS Number:
Molecular Weight:
213.89
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
EC Number:
232-197-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioUltra

Quality Segment

assay

≥99.5% (RT)

form

crystals

impurities

insoluble matter, passes filter test

loss

≤0.1% loss on drying, 20 °C (HV)

color

white to light yellow

pH

3.5-5.5 (25 °C, 0.5 M in H2O)

mp

270  °C ((518 °F ) at 1,013 hPa - OECD Test Guideline 102 -)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

bromide, bromate, chloride, chlorate (as Cl-): ≤100 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

absorption

cut-off at 350 nm in H2O at 0.05 M

SMILES string

[Na+].[O-]I(=O)(=O)=O

InChI

1S/HIO4.Na/c2-1(3,4)5;/h(H,2,3,4,5);/q;+1/p-1

InChI key

JQWHASGSAFIOCM-UHFFFAOYSA-M

Application

Sodium periodate is use in molecular biology and carbohydrate research to cleave carbon-carbon bonds of carbohydrates alginates, chitosans, hyaluronan, scleroglucan/schizophyllan, cellulose nucleic acids other molecules rich in hydroxyls and ketones to prepare them for chemical and physical modifications.
Useful reagent for the oxidation of carbohydrates prior to labeling with biotin hydrazide

Other Notes

Periodate oxidation of agarose gels in enzyme immobilization[1]; Preparation of oligonucleotide 3′-dialcohols in ribonucleoside labeling[2]; Cleavage of cross-linked proteins[3].

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S18783114481.06596
assay

≥99.5% (RT)

assay

≥99.0%

assay

≥99.8%, 99.8-100.3% dry basis (ACS specification)

assay

-

solubility

H2O: 0.5 M at 20 °C, clear, colorless

solubility

H2O: 50 mg/mL

solubility

-

solubility

-

Quality Level

200

Quality Level

300

Quality Level

200

Quality Level

200

form

crystals

form

powder

form

crystalline powder

form

-

mp

270  °C ((518 °F ) at 1,013 hPa - OECD Test Guideline 102 -)

mp

-

mp

300 °C (dec.) (lit.)

mp

300 °C (dec.) (lit.)

cation traces

Al: ≤5 mg/kg, Bi: ≤5 mg/kg, Cd: ≤5 mg/kg, Cr: ≤5 mg/kg, Fe: ≤5 mg/kg, Li: ≤5 mg/kg, Mn: ≤5 mg/kg, Ni: ≤5 mg/kg, Sr: ≤5 mg/kg, Ba: ≤5 mg/kg, Co: ≤5 mg/kg, K: ≤50 mg/kg, Mo: ≤5 mg/kg, Zn: ≤5 mg/kg, Ca: ≤10 mg/kg, Mg: ≤5 mg/kg, Cu: ≤5 mg/kg, Pb: ≤5 mg/kg

cation traces

-

cation traces

Mn: ≤3 ppm

cation traces

-


Still not finding the right product?

Explore all of our products under Sodium (meta)periodate


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Ox. Sol. 1 - Skin Corr. 1C - STOT RE 1

target_organs

thymus gland

Storage Class

5.1A - Strongly oxidizing hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
71859-25G04061832855172
71859-100G04061832855165

Questions

Reviews

No rating value

Active Filters