Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-619-9
MDL number:
Specific activity:
~50 U/mg
Pricing and availability is not currently available.
form
powder
Quality Segment
quality
lyophilized
specific activity
~50 U/mg
color
white
storage temp.
2-8°C
InChI
1S/C11H9N3O2.Na/c15-8-4-5-9(10(16)7-8)13-14-11-3-1-2-6-12-11;/h1-7,16H,(H,12,14);/q;+1/b13-9-;
InChI key
QWZUIMCIEOCSJF-CHHCPSLASA-N
General description
Lipase is a carboxylesterase, which belongs to the α/β hydrolase fold family. It consists of a lid domain, hinge domain, and a catalytic triad Ser-His-Asp/Glu. Aspergillus oryzae lipase has a three-dimensional structure is an α/β hydrolase fold similar to that of the esterase enzyme.[1]
Application
Lipase from Aspergillus oryzae has been used as a component of mixed enzyme solution to determine the viable cell numbers in the biofilms formed by Listeria monocytogenes strains.[2]
Biochem/physiol Actions
Lipases are used industrially for the resolution of chiral compounds and the transesterification production of biodiesel.
Lipases catalyze the hydrolysis of triacylglycerols into glycerol and free fatty acids. Aspergillus oryzae lipase might be used in the production of biodiesel from waste cooking oil. It is used in the acetylation of cellulose and ochratoxin A degradation, and asymmetric organic synthesis.[1] Lipase from Aspergillus oryzae is an industrial enzyme that has various uses in food, detergent, and pharmaceutical industries.[1] It is useful in immobilization studies for 1,3-dioleoyl-2-palmitoylglycerol (OPO) synthesis.[3]
Other Notes
1 U corresponds to the amount of enzyme which liberates 1 μmol oleic acid per minute at pH 8.0 and 40°C (triolein, Cat. No. 62314 as substrate)
1 of 1
This Item | |||
|---|---|---|---|
| specific activity ~50 U/mg | specific activity ≥100,000 U/g | specific activity 15-25 U/mg | specific activity ≥2 U/mg |
| form powder | form solution | form lyophilized solid, powder (fine) | form powder |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| color white | color beige, white | color - | color yellow-brown |
| Quality Level 100 | Quality Level 200 | Quality Level 100 | Quality Level 100 |
| quality lyophilized | quality - | quality lyophilized | quality - |
Still not finding the right product?
Explore all of our products under Lipase from Aspergillus oryzae
signalword
Danger
hcodes
pcodes
Hazard Classifications
Resp. Sens. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




