Skip to Content
Merck

Skip To

62170

Linolenic acid

~70% (GC)

Synonym(s):

α-Lnn, cis,cis,cis-9,12,15-Octadecatrienoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2CH=CH)3(CH2)7CO2H
CAS Number:
Molecular Weight:
278.43
UNSPSC Code:
12352211
NACRES:
NA.77
PubChem Substance ID:
EC Number:
207-334-8
Beilstein/REAXYS Number:
1727693
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~70% (GC)

Quality Level

impurities

~25% linoleic acid, ~5% oleic acid

refractive index

n20/D 1.477, n20/D 1.480 (lit.)

bp

230-232 °C/1 mmHg (lit.)

mp

-11 °C (lit.)

density

0.914 g/mL at 25 °C (lit.)

lipid type

omega FAs

storage temp.

2-8°C

SMILES string

CC/C=C\C/C=C\C/C=C\CCCCCCCC(O)=O

InChI

1S/C18H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h3-4,6-7,9-10H,2,5,8,11-17H2,1H3,(H,19,20)/b4-3-,7-6-,10-9-

InChI key

DTOSIQBPPRVQHS-PDBXOOCHSA-N

Biochem/physiol Actions

An ω-3 fatty acid that serves as a precursor to eicosapentaenoic acid (EPA) but not docosahexaenoic acid. Conversion is greater in women than men, and conversely, β-oxidation metabolism is greater in men than women.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
62210L1376L2626
assay

~70% (GC)

assay

70-80% (GC)

assay

≥98%

assay

≥99% (GC)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

bp

230-232 °C/1 mmHg (lit.)

bp

182 °C/3 mmHg (lit.)

bp

229-230 °C/16 mmHg

bp

182 °C/3 mmHg (lit.)

mp

-11 °C (lit.)

mp

-

mp

−5 °C

mp

-

refractive index

n20/D 1.477, n20/D 1.480 (lit.)

refractive index

n20/D 1.470

refractive index

n20/D 1.466

refractive index

n20/D 1.470 (lit.)

Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under Linolenic acid


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



An Pan et al.
The American journal of clinical nutrition, 96(6), 1262-1273 (2012-10-19)
Prior studies of α-linolenic acid (ALA), a plant-derived omega-3 (n-3) fatty acid, and cardiovascular disease (CVD) risk have generated inconsistent results. We conducted a meta-analysis to summarize the evidence regarding the relation of ALA and CVD risk. We searched multiple
Aliza H Stark et al.
Nutrition reviews, 66(6), 326-332 (2008-06-05)
Consumption of omega 3 fatty acids is known to have health benefits. For many years, the importance of the only member of the omega 3 family considered to be essential, alpha-linolenic acid (ALA), has been overlooked. Current research indicates that
Joel A Simon et al.
The American journal of clinical nutrition, 89(5), 1558S-1564S (2009-03-27)
alpha-Linolenic acid (ALA; 18:3n-3) has been associated inconsistently with an increased risk of prostate cancer. Additional studies have become available since the publication of 2 previous meta-analyses. The objective was to review the published data on the relation between ALA



Global Trade Item Number

SKUGTIN
62170-10ML-F04061832701394
62170-50ML-F04061838177117

Questions

Reviews

No rating value

Active Filters