Skip to Content
Merck

Skip To

62035

Poloxamer 407

oxyethylene 71.5-74.9 %

Synonym(s):

Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol), Lutrol F 127

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352104
NACRES:
NA.25
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Level

mol wt

average mol wt 9840 -14600 g/mol (MMMolecular weightMolecular weight
Molecular wei
g)

composition

oxyethylene, 71.5-74.9%

impurities

≤1 ppm ethylene oxide, ≤0.25% ethylene glycol and diethylene glycol, ≤0.4% total ash, ≤0.75% water, ≤2 ppm arsenic traces, ≤20 ppm heavy metals, ≤5 ppm dioxan, ≤5 ppm propylene oxide, 50-125 ppm butylhydroxytoluene

ign. residue

≤0.30%

color

APHA: ≤120, 50/50 in CH3OH

pH

5.0-7.5 (1:40 in H2O)

solubility

10%, clear, colorless

density

1,06 g/cm3 at 25 °C

application(s)

chemicals and industrial polymers

SMILES string

O2C(C2)C.O1CC1

InChI

1S/C3H6O.C2H4O/c1-3-2-4-3;1-2-3-1/h3H,2H2,1H3;1-2H2

InChI key

RVGRUAULSDPKGF-UHFFFAOYSA-N

Application

Poloxamer 407 has been widely used as a thickening agent and gel binder.

Other Notes

The Poloxamer 407 grades are poloxamers from the extensive line of Pluronic® block copolymers.

Legal Information

Pluronic is a registered trademark of BASF

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.37112P2164020P2163920
form

powder

form

solid

form

-

form

-

application(s)

chemicals and industrial polymers

application(s)

liquid formulation
ophthalmics
parenterals
pharma/biopharma processes
pharmaceutical
semi-solid formulation
solid formulation
solubility enhancement

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

Quality Level

100

Quality Level

500

Quality Level

-

Quality Level

-

impurities

≤0.25% ethylene glycol and diethylene glycol, ≤0.75% water, ≤2 ppm arsenic traces, ≤5 ppm dioxan, 50-125 ppm butylhydroxytoluene, ≤0.4% total ash, ≤20 ppm heavy metals, ≤1 ppm ethylene oxide, ≤5 ppm propylene oxide

impurities

-

impurities

-

impurities

-

mol wt

average mol wt 9840 -14600 g/mol (MMMolecular weightMolecular weight
Molecular wei
g)

mol wt

-

mol wt

-

mol wt

-

density

1,06 g/cm3 at 25 °C

density

1.06 g/cm3 at 25 °C

density

-

density

-


Still not finding the right product?

Explore all of our products under Poloxamer 407


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Jianan Shen et al.
Biomaterials, 34(5), 1581-1590 (2012-11-14)
The metastasis of breast cancer is the leading cause of cancer death in women, and the lung is a common location of a secondary tumor that has metastasized from the primary source tumor. In this work, an attempt to simultaneously
Irshad A Wani et al.
Colloids and surfaces. B, Biointerfaces, 101, 243-250 (2012-09-27)
Silver nanoparticles have been synthesized in the inverse microemulsions formed using three different surfactants viz., cetyl-trimethyl ammonium bromide (CTAB), Tergitol and Triton X-100. We have done a systematic study of the effect of the surfactants on the particle size and
Wendell J Lu et al.
Lipids, 47(6), 571-580 (2012-02-03)
Glucagon-like peptide-1 (GLP-1) and glucose-dependent insulinotropic polypeptide (GIP) are incretins produced in the intestine that play a central role in glucose metabolism and insulin secretion. Circulating concentrations of GLP-1 and GIP are low and can be difficult to assay in



Global Trade Item Number

SKUGTIN
62035-1KG04061832701141

Questions

1–5 of 5 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. I plan to dissolve the Poloxamer 407 powder (Sigma, 16758) in sterile distilled water at 4°C overnight and use it as a solution. Could you provide information on the optimal storage temperature and shelf life for the solution form?

    1 answer
    1. There is no information available on the shelf life of Poloxamer 407 in solution. It is typically recommended to store solutions at room temperature.

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  4. How about the Mw of the product?

    1 answer
    1. The average molecular weight of this product is 10000 to 14600 g/mol.

      Helpful?

  5. is it a pharma grade ?

    1 answer
    1. This product is not pharma grade.

      Helpful?

Reviews

No rating value

Active Filters