Skip to Content
Merck

Skip To

61819

L-Leucine

≥99.5% (NT), suitable for NMR, BioUltra

Synonym(s):

(S)-2-Amino-4-methylpentanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
NACRES:
NA.26
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
12352209
EC Number:
200-522-0
MDL number:
Beilstein/REAXYS Number:
1721722
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

L-Leucine, BioUltra, ≥99.5% (NT)

product line

BioUltra

Quality Segment

assay

≥99.5% (NT)

form

powder or crystals

optical activity

[α]20/D +15.5±0.5°, c = 5% in 5 M HCl

technique(s)

NMR: suitable

impurities

insoluble matter, passes filter test, ≤0.3% foreign amino acids

ign. residue

≤0.1%

loss

≤0.1% loss on drying, 20 °C (HV)

color

white

mp

>300 °C (lit.)

solubility

1 M HCl: 0.1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤200 mg/kg, sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, NH4+: ≤100 mg/kg, Na: ≤100 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

0.1 M in 1 M HCl

UV absorption

λ: 260 nm Amax: 0.05, λ: 280 nm Amax: 0.05

SMILES string

CC(C)C[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1

InChI key

ROHFNLRQFUQHCH-YFKPBYRVSA-N

Other Notes

As spacer in isotachophoresis of serum proteins; Growth requirement of various microorganisms

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L891276526W329703
form

powder or crystals

form

powder

form

-

form

-

assay

≥99.5% (NT)

assay

98.5-101.0%

assay

-

assay

99%

technique(s)

NMR: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

product line

BioUltra

product line

-

product line

TraceCERT®

product line

-

impurities

insoluble matter, passes filter test, ≤0.3% foreign amino acids

impurities

endotoxin, tested

impurities

-

impurities

-

mp

>300 °C (lit.)

mp

>300 °C (lit.)

mp

>300 °C (lit.)

mp

>300 °C (lit.)


Still not finding the right product?

Explore all of our products under L-Leucine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters