Skip to Content
Merck

Skip To

61339

D-Lactose monohydrate

≥99% (HPLC), BioUltra

Synonym(s):

β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, Milk sugar

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
UNSPSC Code:
12352201
NACRES:
NA.25
PubChem Substance ID:
EC Number:
200-559-2
Beilstein/REAXYS Number:
3768231
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioUltra

Quality Level

assay

≥99% (HPLC)

form

solid

optical activity

[α]20/D +53±1°, 22 hr, c = 10% in H2O

impurities

insoluble matter, passes filter test

ign. residue

≤0.05% (as SO4)

color

colorless

pH

3.5-6.0 (25 °C, 0.5 M in H2O)

solubility

H2O: 0.5 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Li: ≤5 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

λ

0.5 M in H2O

UV absorption

λ: 260 nm Amax: 0.02, λ: 280 nm Amax: 0.02

SMILES string

O.OC[C@@H](O)[C@@H](O[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O)[C@H](O)[C@@H](O)C=O

InChI

1S/C12H22O11.H2O/c13-1-4(16)7(18)11(5(17)2-14)23-12-10(21)9(20)8(19)6(3-15)22-12;/h1,4-12,14-21H,2-3H2;1H2/t4-,5+,6+,7+,8-,9-,10+,11+,12-;/m0./s1

InChI key

HBDJFVFTHLOSDW-XBLONOLSSA-N

Application

D-Lactose is a disaccharide of D-glucose and D-galactose units found as a major sugar in milk. D-Lactose is used for the culture of lactic acid metabolizing bacteria, lactic acid bacteria. D-Lactose is used to identify and characterized galectins.

Biochem/physiol Actions

Lactose is a dissacharide formed by the condensation of one galactose and one glucose molecule. Lactose is the major sugar in the milk of most species, typically present between 2-8%. The enzyme lactase hydrolyzes lactose to its constituent monosaccharides. In neonates, glucose released via the action of lactase is a major energy source.

Other Notes

Component of the liquid stationary phase in the GLC separation of nitroxylene and xylenol isomers
Sales restrictions may apply
To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
61341L254PHR1024
assay

≥99% (HPLC)

assay

-

assay

-

assay

-

solubility

H2O: 0.5 M at 20 °C, clear, colorless

solubility

-

solubility

-

solubility

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

300

form

solid

form

solid

form

powder

form

-

impurities

insoluble matter, passes filter test

impurities

-

impurities

dextrose, passes test, sucrose, passes test, ≤0.005% insolubles, 4.0-6.0% water

impurities

-

cation traces

Al: ≤5 mg/kg, Ba: ≤5 mg/kg, Ca: ≤10 mg/kg, Co: ≤5 mg/kg, Cu: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mo: ≤5 mg/kg, Ni: ≤5 mg/kg, Sr: ≤5 mg/kg, As: ≤0.1 mg/kg, Cd: ≤5 mg/kg, Fe: ≤5 mg/kg, Mn: ≤5 mg/kg, Pb: ≤5 mg/kg, Bi: ≤5 mg/kg, Li: ≤5 mg/kg, Zn: ≤5 mg/kg, Cr: ≤5 mg/kg, Na: ≤50 mg/kg

cation traces

-

cation traces

Fe: ≤5 ppm, heavy metals: ≤5 ppm (by ICP)

cation traces

-


Still not finding the right product?

Explore all of our products under D-Lactose monohydrate


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Separation behaviour of some isomeric organic compounds on sugars, sugar alcohols and their mixed phases by gas-liquid chromatography.
Ono, A.
Journal of Chromatography A, 197, 251-251 (1980)
J Nijdam et al.
Colloids and surfaces. B, Biointerfaces, 123, 53-60 (2014-09-30)
Segregation of the protein bovine serum albumin (BSA) and lactose in thin aqueous films during drying was investigated by examining the composition of the dried films using inverse micro Raman spectroscopy (IMRS) and X-ray photoelectron spectroscopy (XPS) sputter-depth profiling. The
Daniel Cavanagh et al.
Applied and environmental microbiology, 81(12), 3961-3972 (2015-04-05)
Lactococcus lactis is predominantly associated with dairy fermentations, but evidence suggests that the domesticated organism originated from a plant niche. L. lactis possesses an unusual taxonomic structure whereby strain phenotypes and genotypes often do not correlate, which in turn has



Global Trade Item Number

SKUGTIN
61339-25G04061832699905
61339-100G04061832699868
61339-250G04061832699899

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters