Skip to Content
Merck

Skip To

56798

Micro particles based on silicon dioxide

size: 1.0 μm

Synonym(s):

Silicon dioxide microsphere, Micro silicon (size: 1 μm), Beads based on silicon dioxide, microsize, Silicon dioxide beads

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352119
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

form

aqueous suspension

concentration

5% (solids)

particle size

1.0 μm

Mw/Mn

0.05

application(s)

glass & ceramic
industrial qc
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

General description

Silicon dioxide based microparticles (size: 1 μm) are ideal to identify particle size distribution (PSD) profile of a particle system.
The 1 μm size of the particles is characterized using CPS disc centrifuge DC 24000.

Application

Silicon dioxide beads, 1.0 μm have a wide range of uses from use as a flow agent to electrical and medical applications.
Used for calibration and regular monitoring of particle size analyzers and surface scanning instruments.
Also used to prepare glass substrates with beads for perovskite-based photovoltaic applications.

Features and Benefits

  • suitable for routine instrument calibration checks, testing and corrections
  • particle size calibrated according to NIST and SRM standards
  • available in 5 mL and 10 mL pack sizes as neat samples


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
811084405456796
particle size

1.0 μm

particle size

2 μm std dev ≤0.2 μm

particle size

5 μm std dev ≤0.35 μm

particle size

0.5 μm

format

neat

format

neat

format

neat

format

neat

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

application(s)

glass & ceramic
industrial qc
pharmaceutical

application(s)

cosmetics
environmental
food and beverages
pharmaceutical

application(s)

glass & ceramic
industrial qc
pharmaceutical

application(s)

glass & ceramic
industrial qc
pharmaceutical

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

form

aqueous suspension

form

aqueous suspension

form

aqueous suspension

form

aqueous suspension


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Digitally-patterned nanoprobe arrays for single cell insertion enabled by wet tapping
Seo HY, et al.
Royal Society of Chemistry Advances, 4(32), 16655-16661 (2014)
Engineering surface atomic structure of single-crystal cobalt (II) oxide nanorods for superior electrocatalysis
Ling T, et al.
Nature Communications, 7(1), 1-8 (2016)
Surachate Kalasin et al.
Langmuir : the ACS journal of surfaces and colloids, 26(4), 2317-2324 (2010-02-10)
This paper explores the particle-level dynamics involved in the capture of gently flowing microparticles on adhesive planar surfaces, governed by electrostatic interactions. The work focuses on conditions which produce sustained microparticle rolling, useful for the development of microfluidic devices which



Global Trade Item Number

SKUGTIN
P055000004061833813744
P6402-5G04061832092775
BP42204061834337324
Y000104504061833805800
PHR3079-500MG04065266900033
P6402-1G04061834393320
1511000-200MG04061833853221
56798-5ML-F04061837790713
56798-10ML-F04061832589589

Questions

Reviews

No rating value

Active Filters