Skip to Content
Merck

Skip To

48760

Gentamicin sulfate from Micromonospora purpurea

≥590 μg/mg of gentamicin

Synonym(s):

Gentamicin C1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
C19-21H39-43N5O7 · 2.5H2SO4
CAS Number:
PubChem Substance ID:
eCl@ss:
42020823
UNSPSC Code:
51281651
NACRES:
NA.76
EC Number:
215-778-9
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Micromonospora purpurea

Quality Segment

form

powder

optical activity

[α]/D 107.0 to 121.0°, c = 10% in H2O

specific activity

≥590 μg/mg of gentamicin

mol wt

Mr 694-723

loss

≤18.0% loss on drying

color

white to off-white

pH

3.5-5.5 (4% in H2O)

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, mycoplasma

mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

OS(O)(=O)=O.CN[C@@H]1[C@@H](O)[C@H](OC[C@]1(C)O)O[C@H]2[C@H](N)C[C@H](N)[C@@H](O[C@H]3O[C@H](CN)CC[C@H]3N)[C@@H]2O.CN[C@@H]4[C@@H](O)[C@H](OC[C@]4(C)O)O[C@H]5[C@H](N)C[C@H](N)[C@@H](O[C@H]6O[C@@H](CC[C@H]6N)[C@@H](C)N)[C@@H]5O.CN[C@H](C)[C@@H]7CC[C@@H](N)[C@H](O7)O[C@@H]8[C@@H](N)C[C@@H](N)[C@H](O[C@H]9OC[C@](C)(O)[C@H](NC)[C@H]9O)[C@H]8O

InChI

1S/C21H43N5O7.C20H41N5O7.C19H39N5O7.H2O4S/c1-9(25-3)13-6-5-10(22)19(31-13)32-16-11(23)7-12(24)17(14(16)27)33-20-15(28)18(26-4)21(2,29)8-30-20;1-8(21)12-5-4-9(22)18(30-12)31-15-10(23)6-11(24)16(13(15)26)32-19-14(27)17(25-3)20(2,28)7-29-19;1-19(27)7-28-18(13(26)16(19)24-2)31-15-11(23)5-10(22)14(12(15)25)30-17-9(21)4-3-8(6-20)29-17;1-5(2,3)4/h9-20,25-29H,5-8,22-24H2,1-4H3;8-19,25-28H,4-7,21-24H2,1-3H3;8-18,24-27H,3-7,20-23H2,1-2H3;(H2,1,2,3,4)/t9-,10-,11+,12-,13+,14+,15-,16-,17+,18-,19-,20-,21+;8-,9-,10+,11-,12+,13+,14-,15-,16+,17-,18-,19-,20+;8-,9+,10-,11+,12-,13+,14+,15-,16+,17+,18+,19-;/m110./s1

InChI key

RDEIXVOBVLKYNT-HDZPSJEVSA-N

General description

Chemical structure: aminoglycoside

Application

Used as a selection agent (gentamicin-resistance gene) in molecular biology applications. It inhibits the growth of a wide variety of grampositive and gram-negative microorganisms including strains resistant to tetracycline, chloramphenicol, kanamycin and colistin, particularly strains of Pseudomonas, Proteus, Staphylococcus and Streptococcus[1].

Biochem/physiol Actions

Mode of Action: Inhibits protein synthesis by binding to L6 protein of 50S ribosomal subunit.
Antimicrobial spectrum: Gram-negative and Gram-positive bacteria, and mycoplasma.

Packaging

1g, 5g, 25g

Preparation Note

Freely soluble in water, practically insoluble in alcohol and ether.

Other Notes

1 U corresponds to 1 μg of the USP gentamicin sulfate reference standard
Keep container tightly closed in a dry and well-ventilated place. Keep in a dry place.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G1272G1522G3632
antibiotic activity spectrum

Gram-negative bacteria, mycoplasma, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, mycoplasma

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, mycoplasma

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, mycoplasma

Quality Level

200

Quality Level

300

Quality Level

300

Quality Level

200

form

powder

form

liquid

form

liquid

form

powder

biological source

Micromonospora purpurea

biological source

microbial

biological source

-

biological source

Micromonospora purpurea

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

color

white to off-white

color

colorless to light yellow

color

colorless to light yellow

color

white to off-white


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
48760-5G-F04061833348536
48760-1G-F04061832393087
48760-25G-F04061832393094

Questions

Reviews

No rating value

Active Filters