Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C4H7NaO3
CAS Number:
Molecular Weight:
126.09
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Pricing and availability is not currently available.
Quality Level
assay
≥96.0% (GC), ≥96.0%
form
powder
composition
sodium, 17.5-19.0%
storage temp.
2-8°C
SMILES string
[Na+].CC(CO)C([O-])=O
InChI
1S/C4H8O3.Na/c1-3(2-5)4(6)7;/h3,5H,2H2,1H3,(H,6,7);/q;+1/p-1
InChI key
RBJZIQZDAZLXEK-UHFFFAOYSA-M
Application
3-Hydroxyisobutyrate (HIBA) may be used to study the distribution, characterization and kinetics of enzymes involved in branched chain amino acid metabolism such as 3-hydroxyisobutyrate dehydrogenase (EC: 1.1.1.31) and 3-hydroxyisobutyryl-CoA hydrolase (EC: 3.1.2.4).
Biochem/physiol Actions
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
1 of 1
This Item | |||
|---|---|---|---|
| assay ≥96.0% (GC) | assay ≥96.0% (GC) | assay ≥98.0% (HPLC) | assay 97.0-103.0% (NT) |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C |
| composition sodium, 17.5-19.0% | composition sodium, 17.5-19.0% | composition - | composition sulfur content, 17.5-18.5% |
| form powder | form - | form powder | form solid |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mammalian 3-hydroxyisobutyrate dehydrogenase.
J W Hawes et al.
Methods in enzymology, 324, 218-228 (2000-09-16)
Disorders of the metabolism of amino acids and related compounds.
Shih, V. E. and Mandell, R. et al.
Genetic Disorders and the Fetus, 514-553 (2010)
Norbert W Lutz et al.
Biochemical and biophysical research communications, 354(1), 160-164 (2007-01-16)
(1)H NMR spectroscopy of cerebrospinal fluid (CSF) is currently being used to study metabolic profiles characteristic of distinct multiple sclerosis (MS) manifestations. For select MS patient groups, we have previously detected significantly increased concentrations of several identified metabolites and one
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 36105-250MG | 04061832404189 |
| 36105-1G | 04061831824025 |



