Skip to Content
Merck
All Photos(1)

Documents

30070

Sigma-Aldrich

Cysteamine

≥98.0% (RT)

Synonym(s):

β-Mercaptoethylamine, 2-Aminoethanethiol, 2-Mercaptoethylamine, Decarboxycysteine, MEA, Thioethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2SH
CAS Number:
Molecular Weight:
77.15
Beilstein:
635649
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (RT)

storage temp.

2-8°C

SMILES string

NCCS

InChI

1S/C2H7NS/c3-1-2-4/h4H,1-3H2

InChI key

UFULAYFCSOUIOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cysteamine is suitable for use:
  • in the preparation of cysteamine modified gold nanoparticles (AuNP)
  • in the fabrication of SU-8 microrods, where in, the amine group of cysteamine reacts with the unreacted epoxide rings present on the surface of the particles, thereby opening it and forming a covalent bond
  • to enhance in vitro development of porcine oocytes matured and fertilized in vitro
  • in a study to demonstrate the depletion effect of cysteamine on cystinotic leucocyte granular fractions of cystine by disulphide interchange
  • as a radioprotector
  • to administer subcutaneously in rats to study its blocking effect on somatostatin secretion without modifying the pancreatic insulin or glucagon content
  • as a scavenger in electrophoretic gels (acetic acid/urea gels)

Biochem/physiol Actions

Cysteamine (β-mercaptoethylamine) depletes cystine from patient′s cells and there by regulates renal glomerular function and increases growth in them. Therefore, cysteamine is considered to be a potential therapeutic for nephropathic cystinosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W A Gahl et al.
The Biochemical journal, 228(3), 545-550 (1985-06-15)
Cystinotic lysosome-rich leucocyte granular fractions, loaded with [35S]cystine, were exposed to different cystine-depleting agents. During a 30 min incubation at 37 degrees C, untreated cystinotic granular fractions lost negligible [35S]cystine when corrected for lysosome rupture. Granular fractions exposed to 0.1
Successful treatment of severe paracetamol overdosage with cysteamine.
L F Prescott et al.
Lancet (London, England), 1(7858), 588-592 (1974-04-06)
R L Sorenson et al.
Diabetes, 32(4), 377-379 (1983-04-01)
Cysteamine (300 mg/kg) administered subcutaneously depletes pancreatic somatostatin to 36% of control levels, but does not alter pancreatic insulin or glucagon content. Although perfusion of pancreata from normal animals with glucose (300 mg/dl) markedly stimulated somatostatin release, pancreata from cysteamine-treated
Cysteamine therapy for children with nephropathic cystinosis
Gahl WA, et al.
The New England Journal of Medicine, 316(16), 971-977 (2018)
C G Grupen et al.
Biology of reproduction, 53(1), 173-178 (1995-07-01)
Most porcine oocytes matured and fertilized in vitro fail to develop normally due to abnormal fertilization. The aim of this study was to determine the effect of cysteamine on pronuclear formation and developmental competence in pig embryos produced in vitro.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service