Skip to Content
Merck

Skip To

261M-9

Glypican-3 (1G12) Mouse Monoclonal Antibody

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
1G12, monoclonal
Application:
IHC (p)
Citations:
7
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

1G12, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (261M-94), vial of 0.5 mL concentrate (261M-95), bottle of 1.0 mL predilute (261M-97), vial of 1.0 mL concentrate (261M-96), bottle of 7.0 mL predilute (261M-98)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1

control

hepatocellular carcinoma (HCC)

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... GPC3(2719)

General description

Glypican-3 (GPC3) is a membrane-bound heparin sulfate proteoglycan known to participate in cell growth and differentiation.[1] GPC3 expression has been observed in the majority of hepatocellular carcinomas (HCC), but is rarely expressed in non-neoplastic hepatic tissue, making it a useful marker for HCC.[1] Aditionally, this marker is expressed in many yolk sac tumors.[1]

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Analysis Note


IVD

IVD

IVD

RUO

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]
Glypican-3 Positive Control Slides, Product No. 261S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5500115264M-9G1921
clone

1G12, monoclonal

clone

SP86, monoclonal

clone

OCH1E5, monoclonal

clone

-

biological source

mouse

biological source

rabbit

biological source

mouse

biological source

human

antibody form

culture supernatant

antibody form

affinity isolated antibody

antibody form

culture supernatant

antibody form

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

-

species reactivity

human

species reactivity

human (tested)

species reactivity

human

species reactivity

-

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

technique(s)

flow cytometry: 1:100, immunoblotting: 1:25, immunohistochemistry: 1:100

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

technique(s)

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Dina H Kandil et al.
Advances in anatomic pathology, 16(2), 125-129 (2009-06-25)
Glypican-3 (GPC3) is a heparan sulfate proteoglycan that plays an important role in cell growth and differentiation. GPC3 function is tissue dependent. In some tissues, GPC3 acts as a tumor suppressor gene, whereas in others, it acts as an oncofetal
Debra L Zynger et al.
The American journal of surgical pathology, 30(12), 1570-1575 (2006-11-24)
Glypican 3 (GPC3), a membrane-bound heparin sulfate proteoglycan, may play a role in promoting embryonic cell growth and differentiation. GPC3 is mutated in Simpson-Golabi-Behmel syndrome, characterized by tissue overgrowth and an increased risk of embryonal malignancies. Recently, GPC3 was reported
Mariana Capurro et al.
Gastroenterology, 125(1), 89-97 (2003-07-10)
Early detection of hepatocellular carcinoma (HCC) is critical for successful treatment. However, the differential diagnosis between HCC and benign hepatic lesions is sometimes difficult and new biochemical markers for HCC are required. It has been reported that glypican-3 (GPC3) messenger



Global Trade Item Number

SKUGTIN
261M-980841683131373
261M-950841683131342
261M-970841683131366
261M-940841683131335
261M-92-VENT04061838384454
261M-960841683131359

Questions

Reviews

No rating value

Active Filters