Skip to Content
Merck

Skip To

21750

Capsaicin

from Capsicum sp., ≥50% (HPLC)

Synonym(s):

8-Methyl-N-vanillyl-trans-6-nonenamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2CHCH=CH(CH2)4CONHCH2C6H3-4-(OH)-3-(OCH3)
CAS Number:
Molecular Weight:
305.41
UNSPSC Code:
12352200
NACRES:
NA.25
PubChem Substance ID:
EC Number:
206-969-8
Beilstein/REAXYS Number:
2816484
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Capsicum sp.

Quality Segment

form

powder

concentration

≥50% (HPLC)

impurities

~35% dihydrocapsaicin

mp

62-65 °C (lit.), 62-66 °C

solubility

H2O: insoluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

COc1cc(CNC(=O)CCCC\C=C\C(C)C)ccc1O

InChI

1S/C18H27NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h6,8,10-12,14,20H,4-5,7,9,13H2,1-3H3,(H,19,21)/b8-6+

InChI key

YKPUWZUDDOIDPM-SOFGYWHQSA-N

Gene Information

General description

Capsaicin occurs as the active ingredient of hot/red pepper and was first obtained by Thresh in 1846. It is a lipophilic vanilloid compound responsible for the acrid taste of hot peppers.[1][2][3]

Application

Capsaicin has been used in the development and pharmaceutical production of a gastrointestinal mucosal protective drug.

Biochem/physiol Actions

Capsaicin shows its activity by binding to vanilloid receptors and eliciting a nociceptive response. It shows an analgesic effect in neuropathic and musculoskeletal disorders. Capsaicin is also used in the management of bladder detrusor hyperreflexia.[1][2][4][3][5]
Prototype vanilloid receptor agonist. Neurotoxin; activates sensory neurons that give rise to unmyelinated C-fibers, many of which contain substance P. Topical application desensitizes the sensory nerve endings giving a paradoxical antinociceptive effect; systemic administration can be neurotoxic to capsaicin-sensitive cells, especially in newborn animals. Active component of chili peppers.
Prototype vanilloid receptor agonist; neurotoxin. Active component of chili peppers.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M2028360376PHR1450
biological source

Capsicum sp.

biological source

Capsicum sp.

biological source

-

biological source

-

form

powder

form

-

form

powder

form

-

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

300

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

application(s)

-

application(s)

-

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

H2O: insoluble

solubility

H2O: insoluble, ethanol: soluble

solubility

chloroform: 5%

solubility

-


Still not finding the right product?

Explore all of our products under Capsaicin


signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



M J Caterina et al.
Science (New York, N.Y.), 288(5464), 306-313 (2000-04-15)
The capsaicin (vanilloid) receptor VR1 is a cation channel expressed by primary sensory neurons of the "pain" pathway. Heterologously expressed VR1 can be activated by vanilloid compounds, protons, or heat (>43 degrees C), but whether this channel contributes to chemical
Capsaicin is a New Gastrointestinal Mucosal Protecting Drug Candidate in Humans?Pharmaceutical Development and Production Based on Clinical Pharmacology
Capsaicin-sensitive Neural Afferentation and the Gastrointestinal Tract: From Bench to Bedside (2014)
Lorna Mason et al.
BMJ (Clinical research ed.), 328(7446), 991-991 (2004-03-23)
To determine the efficacy and safety of topically applied capsaicin for chronic pain from neuropathic or musculoskeletal disorders. Cochrane Library, Medline, Embase, PubMed, an in-house database, and contact with manufacturers of topical capsaicin. Randomised controlled trials comparing topically applied capsaicin



Global Trade Item Number

SKUGTIN
21750-5G-F04061838062413
21750-100MG-F04061838774927
21750-1G-F04061838062406

Questions

Reviews

No rating value

Active Filters