Skip to Content
Merck

Skip To

05019

7-Diethylamino-3-[N-(2-maleimidoethyl)carbamoyl]coumarin

BioReagent, suitable for fluorescence, ≥97.0% (HPLC)

Synonym(s):

MDCC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C20H21N3O5
CAS Number:
Molecular Weight:
383.40
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Level

assay

≥97.0% (HPLC)

suitability

suitable for fluorescence

SMILES string

CCN(CC)c1ccc2C=C(C(=O)NCCN3C(=O)C=CC3=O)C(=O)Oc2c1

InChI

1S/C20H21N3O5/c1-3-22(4-2)14-6-5-13-11-15(20(27)28-16(13)12-14)19(26)21-9-10-23-17(24)7-8-18(23)25/h5-8,11-12H,3-4,9-10H2,1-2H3,(H,21,26)

InChI key

IXQPRUQVJIJUEB-UHFFFAOYSA-N

Application

Thiol-reactive probe for protein labelling
7-Diethylamino-3-[N-(2-maleimidoethyl)carbamoyl]coumarin is utilized as a fluoresencent biological sensing device . Used for real-time measurements for the release of inorganic phosphates during enzymatic reaction when MDCC is conjugated to a mutant phosphate-binding protein . Also, utilized for intramolecular fluorescence energy transfer (FRET) experiments .

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G762700062107M9307
suitability

suitable for fluorescence

suitability

-

suitability

-

suitability

-

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

200

assay

≥97.0% (HPLC)

assay

≥90%

assay

-

assay

-

product line

BioReagent

product line

-

product line

-

product line

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Lyndon L E Salins et al.
Analytical and bioanalytical chemistry, 372(1), 174-180 (2002-04-10)
A sensing system for nickel based on the nickel binding protein (NBP) from Escherichia coli is shown to be feasible. The versatility of NBP was demonstrated by its use in three different assay formats. When the NBP binds nickel, it
M Brune et al.
Biochemistry, 33(27), 8262-8271 (1994-07-12)
A probe has been developed that can rapidly measure micromolar concentrations of inorganic phosphate (Pi), in particular to follow the release of Pi in real time from enzymes such as phosphatases. Its application is described to investigate the mechanism of
Yu-Chih Tsai et al.
Biochemistry, 45(32), 9675-9687 (2006-08-09)
We show that T7 DNA polymerase exists in three distinct structural states, as reported by a conformationally sensitive fluorophore attached to the recognition (fingers) domain. The conformational change induced by a correct nucleotide commits the substrate to the forward reaction



Global Trade Item Number

SKUGTIN
1483185-10MG04061833852651
150215-5G04061831818727
PHR3071-25MG04065266899955
150215-1G04061825583037
05019-1MG-F04061823353243
05019-5MG-F04061833488553

Questions

  1. What do the excitation/emission peaks of this dye look like?

    1 answer
    1. The product specification requires an emission wavelength range of 460-470nm. The product is not screened for excitation wavelength, however it is expected to fall within a range of 416 to 425 nm. The compound is typically used for real-time measurements for the release of inorganic phosphates during enzymatic reaction when MDCC is conjugated to a mutant phosphate-binding protein . Please see the link below to review a publication that may be of interest:
      https://pubs.acs.org/doi/10.1021/bi9804277

      Helpful?

Reviews

No rating value

Active Filters