Skip to Content
Merck

Skip To

E7750

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride

commercial grade, powder

Synonym(s):

N-Ethyl-N′-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDAC, EDC, EDC hydrochloride, WSC hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H17N3 · HCl
CAS Number:
Molecular Weight:
191.70
UNSPSC Code:
12352111
NACRES:
NA.31
PubChem Substance ID:
EC Number:
247-361-2
Beilstein/REAXYS Number:
5764110
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

technique(s)

Northern blotting: suitable, bioconjugation: suitable

color

white to off-white

mp

110-115 °C (lit.)

solubility

H2O: ≤100 mg/mL

storage temp.

−20°C

SMILES string

Cl.CCN=C=NCCCN(C)C

InChI

1S/C8H17N3.ClH/c1-4-9-8-10-6-5-7-11(2)3;/h4-7H2,1-3H3;1H

InChI key

FPQQSJJWHUJYPU-UHFFFAOYSA-N

General description

1-Ethyl-3-(3-dimethylaminopropyl) carbodiimide hydrochloride (EDC HCl), commonly referred to as EDAC, is a highly potent and extensively utilized water-soluble reagent in the realm of chemical and biochemical research, playing a crucial role in facilitating the formation of amide bonds. Within the domain of peptide synthesis, EDC HCl demonstrates notable efficiency by catalyzing the coupling of amino acids through their carboxyl and amine groups, thereby facilitating the creation of peptide backbones with specific sequences and functionalities. This particular attribute is highly valuable in the production of peptides for various research purposes. Expanding beyond peptides, EDC HCl′s influence extends to the construction of immunogens, where it actively participates in the covalent attachment of haptens (small molecules that elicit an immune response) to carrier proteins. This involvement proves instrumental in the research and development of vaccines.

The versatility of EDC HCl further manifests in its capacity to modify nucleic acids, allowing for the labeling of DNA and RNA through their 5′ phosphate groups. This functionality enhances the visualization, tracking, and analysis of these crucial molecules, contributing significantly to the progression of nucleic acid research. Moreover, EDC HCl serves as a vital biomolecule bridge, acting as a crosslinker that connects amine-reactive NHS-esters of biomolecules to carboxyl groups. This technique is particularly valuable in protein conjugation, enabling the creation of hybrid molecules with novel properties and functions. The underlying mechanism of EDC HCl involves its reaction with a carboxyl group, forming an unstable intermediate that actively seeks an amine partner. The delicate balance of this reaction emphasizes the need for optimizing conditions to ensure efficient conjugation. The assistance of N-hydroxysuccinimide (NHS) further enhances EDC HCl′s capabilities by stabilizing the intermediate and enabling two-step conjugation procedures, offering greater flexibility and control, especially when dealing with complex biomolecules.

Application

  • N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride has been used for the formation of FND (fluorescent nanodiamonds)-transferrin bioconjugates.
  • It has been used for crosslinking polyethylenimine to gold particles.[1]
  • It has been used as a carbodiimide linkage agent for coating of carboxylated polystyrene beads with biotinylated BSA (bovine serum albumin).

Biochem/physiol Actions

Water soluble condensing reagent. EDAC is generally utilized as a carboxyl activating agent for amide bonding with primary amines. In addition, it will react with phosphate groups. EDAC has been used in peptide synthesis; crosslinking proteins to nucleic acids; and preparation of immunoconjugates as examples. Typically, EDAC is utilized in the pH range 4.0-6.0 without buffers. In particular, amine and carboxylate buffers should be avoided.

Features and Benefits

Versatile and adaptable for a wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E6383034498.00907
form

powder

form

crystalline

form

powder

form

solid

technique(s)

Northern blotting: suitable, bioconjugation: suitable

technique(s)

-

technique(s)

bioconjugation: suitable

technique(s)

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C (−15°C to −25°C)

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

200

solubility

H2O: ≤100 mg/mL

solubility

H2O: ≤100 mg/mL

solubility

H2O: soluble 0.2 g/L

solubility

-

color

white to off-white

color

white to off-white

color

-

color

-


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Stomach,large intestine,lymph node

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content






Global Trade Item Number

SKUGTIN
E7750-1KG04061835544653
E7750-5KG04061832862132
E7750-100MG04061835544646
E7750-10G04061835516018
E7750-5G04061835538379
E7750-100G04061835544639
E7750-1G04061835483433
E7750-25G04061835544660

Questions

1–7 of 7 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. What impurities might be present in N-(3-Dimethylaminopropyl)-N´-ethylcarbodiimide hydrochloride, Product E7750?

    1 answer
    1. The two compounds most likely to appear in EDAC as trace impurities are 3-dimethylaminopropylamine and N-(3-dimethylaminopropyl)-N'-ethylurea as a result of synthesis (intermediate) and degradation (moisture or coupling reaction), respectively.

      Helpful?

  4. Product No. E7750, N-(3-Dimethylaminopropyl)-N´-ethylcarbodiimide hydrochloride, is listed as "commercial grade". What does that mean?

    1 answer
    1. It means that it is the grade of N-(3-Dimethylaminopropyl)-N´-ethylcarbodiimide (EDAC) hydrochloride that was commercially available at the time we purchased it. This product is not assayed for purity. If you need a high purity EDAC, consider Product No. 03450.

      Helpful?

  5. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  6. Can N-(3-Dimethylaminopropyl)-N´-ethylcarbodiimide hydrochloride, Product E7750, be used as a peptide coupling reagent in alcoholic solvents? If so, do you have a reference?

    1 answer
    1. Yes. Nosaki, S., Effects of amounts of additives on peptide coupling mediated by a water-soluble carbodiimide in alcohols. J. Peptide Res., 54(2), 162-167 (1999). This paper describes using the E7750 along with 3,4-dihydro-3-hydroxy-4-oxo-1,2,3-benzotriazine (HOOBt) and 1-hydroxy-7-azabenzotriazole (HOAt) for enhancement of peptide coupling.

      Helpful?

  7. I understand that E7750, N-(3-Dimethylaminopropyl)-N´-ethylcarbodiimide hydrochloride, can be used in peptide synthesis. Do you have a brochure on some other reagents that can be used for peptide synthesis?

    1 answer
    1. Yes.

      Helpful?

Reviews

No rating value

Active Filters