Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Quality Segment
agency
USP/NF, meets USP testing specifications
form
solid
mp
191-192 °C (lit.)
storage temp.
2-8°C
SMILES string
NC(=O)N1c2ccccc2C=Cc3ccccc13
InChI
1S/C15H12N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-10H,(H2,16,18)
InChI key
FFGPTBGBLSHEPO-UHFFFAOYSA-N
Gene Information
human ... PRNP(5621), SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN7A(6332), SCN8A(6334), SCN9A(6335)
rat ... Scn2a1(24766), Scnn1a(25122), Scnn1g(24768), Slc6a1(79212)
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| form solid | form - | form - | form - |
| Quality Level 200 | Quality Level 300 | Quality Level - | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-30°C | storage temp. - | storage temp. 2-8°C |
| Gene Information human ... PRNP(5621), SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN7A(6332), SCN8A(6334), SCN9A(6335) | Gene Information human ... SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN7A(6332), SCN8A(6334), SCN9A(6335) | Gene Information human ... SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN7A(6332), SCN8A(6334), SCN9A(6335) | Gene Information human ... SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN7A(6332), SCN8A(6334), SCN9A(6335) |
| agency USP/NF, meets USP testing specifications | agency traceable to Ph. Eur. C0450000, traceable to USP 1093001 | agency - | agency - |
| mp 191-192 °C (lit.) | mp 191-192 °C (lit.) | mp 191-192 °C (lit.) | mp 191-192 °C (lit.) |
Still not finding the right product?
Explore all of our products under Carbamazepine
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Repr. 1A - Resp. Sens. 1 - Skin Sens. 1A - STOT SE 3
target_organs
Central nervous system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C8981-25G | 04061833523070 |
| C8981-5G | 04061833523087 |


