Skip to Content
Merck

Skip To

C3000000

Cyanocobalamin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Vitamin B12, α-(5,6-Dimethylbenzimidazolyl)cyanocobamide, CN-Cbl, Cyanocob(III)alamin, Cyanocobalamin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C63H88CoN14O14P
CAS Number:
Molecular Weight:
1355.37
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Beilstein/REAXYS Number:
4122889
eCl@ss:
34058004
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

grade

pharmaceutical primary standard

agency

EP

API family

cyanocobalamin

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

mp

>300 °C (decomposes before melting)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

C/C(C1=N[C@]([C@]([C@H](CC(N)=O)[C@@]/2(C)CCC(NC[C@@H](OP([O-])(O[C@H]3[C@@H](O)[C@@H](N4C=[N]5C6=C4C=C(C)C(C)=C6)O[C@@H]3CO)=O)C)=O)([H])N([Co+]5C#N)C2=C(C)/C([C@@H](CCC(N)=O)C/7(C)C)=NC7=C/8)(C)[C@@](C)(CC(N)=O)[C@@H]1CCC(N)=O)=C9N=C8[C@@H](CCC(N)=O)[C@

InChI

1S/C62H90N13O14P.CN.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;1-2;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57?,59-,60+,61+,62+;;/m1../s1

InChI key

RMRCNWBMXRMIRW-QJRSUKKJSA-L

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Cyanocobalamin, a B vitamin with potential applications in medical and food industries, is vital to growth, cell reproduction, hematopoiesis, etc. It is utilized for the management of trigeminal neuralgia and multiple sclerosis.

Application

Cyanocobalamin may be used as an EP reference standard for the determination of the analyte in pharmaceutical formulations by various chromatography techniques.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1234BP466C3607
application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)
vitamins, nutraceuticals, and natural products

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

-

format

neat

format

neat

format

neat

format

-

form

solid

form

-

form

solid

form

powder

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Still not finding the right product?

Explore all of our products under Cyanocobalamin


Storage Class

11 - Combustible Solids

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ultra-high performance liquid chromatographic and mass spectrometric analysis of active vitamin B12 in cells of Propionibacterium and fermented cereal matrices.
Chamlagain B, Edelmann M, Kariluoto S, et al.
Inflammation and regeneration, 166, 630-638 (2014)
LC simultaneous determination of Thioctic Acid, Benfotiamine and Cyanocobalamin in Thiotacid Compound capsules
Salem H
Chromatographia, 72(3-4), 327-330 (2010)
Microbial production of vitamin B 12: a review and future perspectives
Fang, Huan and Kang, Jie and Zhang, Dawei
Microbial cell factories, 16(1), 15-15 (2017)



Global Trade Item Number

SKUGTIN
C300000004061833792599

Questions

Reviews

No rating value

Active Filters