Skip to Content
Merck

Skip To

95437

L-Cysteine

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(R)-2-Amino-3-mercaptopropionic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
HSCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
121.16
E Number:
E920
PubChem Substance ID:
eCl@ss:
32160406
UNSPSC Code:
41116107
NACRES:
NA.24
EC Number:
200-158-2
MDL number:
Beilstein/REAXYS Number:
1721408
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

TraceCERT®

grade

certified reference material, TraceCERT®

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

amino acids, peptides, proteins

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

240 °C (dec.) (lit.)

UV absorption

λ: 260 nm Amax: 1.5

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

N[C@@H](CS)C(O)=O

InChI

1S/C3H7NO2S/c4-2(1-7)3(5)6/h2,7H,1,4H2,(H,5,6)/t2-/m0/s1

InChI key

XUJNEKJLAYXESH-REOHCLBHSA-N

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.


Check out the entire portfolio of our amino acids certified reference materials (CRM)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

NMDA glutamatergic receptor agonist.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
C7352778672W326305
application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

-

application(s)

-

application(s)

flavors and fragrances

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

-

technique(s)

-

format

neat

format

-

format

-

format

-

Quality Level

300

Quality Level

200

Quality Level

100

Quality Level

300

grade

certified reference material, TraceCERT®

grade

-

grade

-

grade

FG, Halal

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

-


Still not finding the right product?

Explore all of our products under L-Cysteine


flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

11 - Combustible Solids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters