Skip to Content
Merck

Skip To

83059

M-Gredient® Potato Peptone from potatoes

suitable for microbiology

Synonym(s):

Peptone from vegetable

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

potato

Quality Segment

form

powder

composition

amino-nitrogen, ≥4.5% , total nitrogen (N), ≥9.5%

ign. residue

≤22%

loss

≤6.0% loss on drying

pH

6.5-7.5 (25 °C)

solubility

H2O: 2%, slightly turbid to turbid, yellow to brownish-yellow

application(s)

food and beverages, microbiology

Application

Manufactured by a controlled enzymatic hydrolysis of potato proteins. Source of organic nitrogen and growth factors recommended in media for industrial fermentation.

Legal Information

M-GREDIENT is a registered trademark of Merch KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
381439297696174
biological source

potato

biological source

potato

biological source

plant

biological source

plant

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

food and beverages, microbiology

application(s)

microbiology

application(s)

food and beverages
vitamins, nutraceuticals, and natural products, microbiology

application(s)

food and beverages
microbiology

form

powder

form

powder

form

powder

form

powder

pH

6.5-7.5 (25 °C)

pH

6.7-7.7 (25 °C, 2% in H2O)

pH

6.6±0.5 (25 °C, 2% in H2O)

pH

6.7-7.7

solubility

H2O: 2%, slightly turbid to turbid, yellow to brownish-yellow

solubility

H2O: 2%, clear (light yellow and light brown-yellow and light brown)

solubility

H2O: 2%, clear, light brownish-yellow

solubility

-


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
83059-10G-KC-F04061832573427
83059-500G-F04061833005408

Questions

Reviews

No rating value

Active Filters