Skip to Content
Merck

Skip To

75423

Vancomycin supplement

suitable for microbiology

Synonym(s):

Vancomycin hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C66H75Cl2N9O24 · xHCl
CAS Number:
Molecular Weight:
1449.25 (free base basis)
MDL number:
UNSPSC Code:
41171614
NACRES:
NA.85
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

according to GB 4789.40-2016, according to ISO 22964:2017

sterility

sterile

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages, microbiology

storage temp.

2-8°C

suitability

Enterobacter spp.

SMILES string

[Cl-].Clc1c2ccc(c1)[C@H]([C@@H]3NC(=O)[C@H](NC(=O)[C@@H]6NC(=O)[C@@H](NC(=O)[C@@H]([C@@H](c7cc(c(cc7)Oc8c(c(cc6c8)O2)O[C@@H]9O[C@@H]([C@H]([C@@H]([C@H]9O[C@@H]%10O[C@H]([C@H]([C@](C%10)(N)C)O)C)O)O)CO)Cl)O)NC(=O)[C@H]([N+H2]C)CC(C)C)CC(=O)N)c4cc(c(cc4)O)c

InChI

1S/C66H75Cl2N9O24.ClH/c1-23(2)12-34(71-5)58(88)76-49-51(83)26-7-10-38(32(67)14-26)97-40-16-28-17-41(55(40)101-65-56(54(86)53(85)42(22-78)99-65)100-44-21-66(4,70)57(87)24(3)96-44)98-39-11-8-27(15-33(39)68)52(84)50-63(93)75-48(64(94)95)31-18-29(79)19-37(81)45(31)30-13-25(6-9-36(30)80)46(60(90)77-50)74-61(91)47(28)73-59(89)35(20-43(69)82)72-62(49)92;/h6-11,13-19,23-24,34-35,42,44,46-54,56-57,65,71,78-81,83-87H,12,20-22,70H2,1-5H3,(H2,69,82)(H,72,92)(H,73,89)(H,74,91)(H,75,93)(H,76,88)(H,77,90)(H,94,95);1H/t24-,34+,35-,42+,44-,46+,47+,48-,49+,50-,51+,52+,53+,54-,56+,57+,65-,66-;/m0./s1

InChI key

LCTORFDMHNKUSG-XTTLPDOESA-N

Application

Vancomycin supplement is an antibiotic supplement recommended by ISO Committee for selective enrichment of Enterobacter sakazakii. It has been used as a supplement in a selective media to study the presence of vancomycin resistant enterococci in the treated effluent of wastewater treatment plants.

Other Notes

(per vial, sufficient for 1000 ml medium)
Vancomycin 10.00 mg

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1732V1130V0045000
application(s)

environmental
food and beverages, microbiology

application(s)

pharmaceutical (small molecule)

application(s)

agriculture

application(s)

pharmaceutical (small molecule)

form

powder

form

-

form

powder

form

-

suitability

Enterobacter spp.

suitability

-

suitability

-

suitability

-

sterility

sterile

sterility

-

sterility

-

sterility

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-

storage temp.

2-8°C

storage temp.

-10 to -25°C

storage temp.

2-8°C

storage temp.

−20°C


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters