Skip to Content
Merck

Skip To

68097

(−)-Epicatechin

analytical standard

Synonym(s):

(−)-cis-3,3′,4′,5,7-Pentahydroxyflavane, (2R,3R)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H14O6
CAS Number:
Molecular Weight:
290.27
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
207-710-1
Beilstein/REAXYS Number:
92760
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

240 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

2-8°C

SMILES string

O[C@@H]1Cc2c(O)cc(O)cc2O[C@@H]1c3ccc(O)c(O)c3

InChI

1S/C15H14O6/c16-8-4-11(18)9-6-13(20)15(21-14(9)5-8)7-1-2-10(17)12(19)3-7/h1-5,13,15-20H,6H2/t13-,15-/m1/s1

InChI key

PFTAWBLQPZVEMU-UKRRQHHQSA-N

General description

(−)-Epicatechin is a monomeric catechin and a polyphenolic component, belonging to the class of flavan-3-ols. It is majorly found in apples, apple products and green tea.

Application

(−)-Epicatechin may be used as an analytical reference standard for the quantification of the analyte in green tea using high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
7487303940590E4018
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

application(s)

food and beverages

application(s)

food and beverages

application(s)

food and beverages

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

format

neat

format

neat

format

-

format

-

Quality Level

100

Quality Level

300

Quality Level

-

Quality Level

200

grade

analytical standard

grade

certified reference material, TraceCERT®

grade

primary reference standard

grade

-

assay

≥97.0% (HPLC)

assay

-

assay

-

assay

≥98% (HPLC)


Still not finding the right product?

Explore all of our products under (−)-Epicatechin


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A method for fast determination of epigallocatechin gallate (EGCG), epicatechin (EC), catechin (C) and caffeine (CAF) in green tea using HPLC
Saito.TS, et al.
Food Sci. Technol., 26(2), 394-400 (2006)
Recent Advances in Polyphenol Research, 26(2), 394-400 (2009)
Alice Cattivelli et al.
Foods (Basel, Switzerland), 10(5) (2021-06-03)
The impact of domestic cooking (baking, boiling, frying and grilling) and in vitro digestion on the stability and release of phenolic compounds from yellow-skinned (YSO) and red-skinned onions (RSO) have been evaluated. The mass spectrometry identification pointed out flavonols as



Global Trade Item Number

SKUGTIN
68097-10MG04061832751054

Questions

Reviews

No rating value

Active Filters