Skip to Content
Merck

Skip To

676802

Ethylene carbonate

anhydrous, 99%

Synonym(s):

1,3-Dioxolan-2-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C3H4O3
CAS Number:
Molecular Weight:
88.06
EC Number:
202-510-0
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
106249
MDL number:
Assay:
99%
Grade:
anhydrous
Bp:
243-244 °C/740 mmHg (lit.)
Vapor pressure:
0.02 mmHg ( 36.4 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

anhydrous

Quality Segment

vapor density

3.04 (vs air)

vapor pressure

0.02 mmHg ( 36.4 °C)

assay

99%

form

solid

expl. lim.

3.6-16.1 % (lit.)

impurities

≤0.006% water

bp

243-244 °C/740 mmHg (lit.)

mp

35-38 °C (lit.)

density

1.321 g/mL at 25 °C (lit.)

SMILES string

O=C1OCCO1

InChI

1S/C3H4O3/c4-3-5-1-2-6-3/h1-2H2

InChI key

KMTRUDSVKNLOMY-UHFFFAOYSA-N

General description

Ethylene carbonate is a cyclic carbonate with high dielectric constant.

Application

Applications of ethylene carbonate (EC):
  • EC can be an effective liquefying reagent for cellulose liquefaction.
  • Dimethyl carbonate (DMC) can be synthesized via the transesterification of EC and methanol.
  • EC can be a non-hazardous alternative to phosgene and isocyanates for the synthesis of polyurethanes.



Still not finding the right product?

Explore all of our products under Ethylene carbonate


pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

target_organs

Kidney

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

289.4 °F - closed cup

flash_point_c

143 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters