Skip to Content
Merck

Skip To

62862

Sodium dodecyl sulfate

greener alternative

≥90% ((Assay))

Synonym(s):

Dodecyl sodium sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, SDS, Sodium lauryl sulfate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)11OSO3Na
CAS Number:
Molecular Weight:
288.38
NACRES:
NA.21
PubChem Substance ID:
eCl@ss:
39093306
UNSPSC Code:
12161900
EC Number:
205-788-1
MDL number:
Beilstein/REAXYS Number:
3599286
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥90% ((Assay))

form

(Rods)

mol wt

288.38 g/mol

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

description

anionic, 62

technique(s)

LC/MS: suitable, electrophoresis: suitable, protein quantification: suitable

mp

204-207 °C (lit.)

solubility

water: soluble

density

(1.03 g/cm3 at 20 °C (68 °F))

HLB

40

application(s)

(Western Blot)

greener alternative category

SMILES string

[Na+].CCCCCCCCCCCCOS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1

InChI key

DBMJMQXJHONAFJ-UHFFFAOYSA-M

General description

Sodium dodecyl sulfate (SDS), also known as sodium lauryl sulfate (SLS), has an alkyl tail of 12 carbon atoms attached to a sulfate group. It is an anionic surfactant due to the negative charge of its sulfate group.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is used as a surfactant to enhance catalysis.  Click here for more information.

Application

Sodium dodecyl sulfate has been used:

  • to prepare Chit for coating the Janus micromotors for the synthesis of magnesium (Mg)-based micromotors[1]
  • as a component in low salt lysis buffer to lyse the edible bird’s nest (EBN) treated cells for western blot analysis[2]
  • in the preparation of decellularizing solution to scale down and decellularize skin fragments
Anionic detergent

Biochem/physiol Actions

Sodium dodecyl sulfate (SDS) is frequently used as an ingredient in a wide range of products, including pharmaceuticals, and product formulations. It is broadly used in several applications including electrophoresis for the separation of proteins and as an organic template for preparing periodic mesoporous organosilica nanospheres. SDS is also used in the solubilization of proteins and lipids, as well as drugs, dispersion of functionalized carbon nanotubes and graphene sheets, and gas hydrate formation. It is commonly used for the quick disruption of biological membranes. Due to its ability to disrupt the tissue architecture, SDS is preferred in several nucleic acid purification reagents. SDS has very low solubility in high salt, and chaotropic solutions and is readily precipitable in the presence of potassium salts, so it is usually not added to guanidinium buffers.

Features and Benefits

  • Assay: ≥90%
  • Suitable for LC/MS, electrophoresis, and protein quantification
  • Inhibits RNase and deoxyribonuclease (DNase) activity

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
L450971726L6026
form

(Rods)

form

powder

form

rod

form

powder

technique(s)

LC/MS: suitable, protein quantification: suitable, electrophoresis: suitable

technique(s)

-

technique(s)

ion pair chromatography: suitable

technique(s)

-

assay

≥90% ((Assay))

assay

≥98.5% (GC)

assay

≥99.0% (GC)

assay

≥99.0% (GC)

application(s)

(Western Blot)

application(s)

advanced drug delivery

application(s)

-

application(s)

-

Quality Level

100

Quality Level

200

Quality Level

100

Quality Level

200

solubility

water: soluble

solubility

water: soluble

solubility

water: soluble

solubility

H2O: 0.1 M, clear, colorless


Still not finding the right product?

Explore all of our products under Sodium dodecyl sulfate


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 2

flash_point_f

338.0 °F

flash_point_c

170 °C

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters