Skip to Content
Merck

Skip To

54969

Clenbuterol-d9 hydrochloride

analytical standard

Synonym(s):

1-(4-Amino-3,5-dichlorophenyl)-2-(tert-butyl-d9-amino)ethanol hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C12D9H9Cl2N2O · HCl
CAS Number:
Molecular Weight:
322.71
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥97.0% (isotopic purity), ≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+9

storage temp.

2-8°C

SMILES string

Cl.[2H]C([2H])([2H])C(NCC(O)c1cc(Cl)c(N)c(Cl)c1)(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C12H18Cl2N2O.ClH/c1-12(2,3)16-6-10(17)7-4-8(13)11(15)9(14)5-7;/h4-5,10,16-17H,6,15H2,1-3H3;1H/i1D3,2D3,3D3;

InChI key

OPXKTCUYRHXSBK-KYRNGWDOSA-N

General description

Clenbuterol-d9 hydrochloride may be used as an internal standard for the quantification of clenbuterol in biological samples by ultra-high pressure liquid chromatography–tandem mass spectrometry (UPLC-MS/MS).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
949723282532143
application(s)

clinical testing

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

application(s)

forensics and toxicology
pharmaceutical (small molecule)

application(s)

forensics and toxicology
veterinary

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable, solid phase extraction (SPE): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

analytical standard

assay

≥97.0% (isotopic purity), ≥98.0% (HPLC)

assay

≥99.0% (HPLC)

assay

-

assay

-

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Quantification of clenbuterol at trace level in human urine by ultra-high pressure liquid chromatography?tandem mass spectrometry.
Nicoli R, et al.
Journal of Chromatography A, 1292, 142-150 (2013)



Global Trade Item Number

SKUGTIN
54969-10MG04061832575407

Questions

Reviews

No rating value

Active Filters